Skip to Content
Merck
All Photos(1)

Key Documents

F3771

Sigma-Aldrich

Fumonisin B2 from Fusarium moniliforme

sphingosine N-acyltransferase inhibitor

Synonym(s):

FB2, Fumonisin B2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H59NO14
CAS Number:
Molecular Weight:
705.83
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

Fusarium sp. (Fusarium moniliforme)

Assay

≥96% (TLC)

form

powder

solubility

methanol: 10 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CCCCC(C)C(OC(=O)CC(CC(O)=O)C(O)=O)C(CC(C)CCCCCCC(O)CC(O)C(C)N)OC(=O)CC(CC(O)=O)C(O)=O

InChI

1S/C34H59NO14/c1-5-6-12-21(3)32(49-31(43)18-24(34(46)47)16-29(40)41)27(48-30(42)17-23(33(44)45)15-28(38)39)14-20(2)11-9-7-8-10-13-25(36)19-26(37)22(4)35/h20-27,32,36-37H,5-19,35H2,1-4H3,(H,38,39)(H,40,41)(H,44,45)(H,46,47)

InChI key

UXDPXZQHTDAXOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fumonisin B2 (FB2) is a diester of tricarballylic acid and polyhydric alcohols.[1]. Fumonisins B2 (FB2) does not have one or more hydroxyl groups at the C5 or C10 positions.[2]
Mycotoxin originally isolated from Fusarium sp, which grows on corn. Produces neural tube defects and craniofacial abnormalities in fetuses. Many strains of Aspergillus niger growing naturally on grapes are capable of producing fumonisin B2, so it is now a concern for human consumption of grapes, raisins, and wine.

Application

Fumonisin B2 (FB2) from Fusarium moniliforme has been used:
  • as a standard for the detection of FB2 by high performance liquid chromatography (HPLC) in corn based food products[1][3]
  • used as standard in the determination of fumonisin B1 and B2 in corn and corn-based products by high-performance liquid chromatography[3]
  • as a standard in the determination of mycotoxins by HPLC[4]

Biochem/physiol Actions

Inhibitor of sphingosine N-acyltransferase. Blocks the formation of ceramide from sphingosine.

related product

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumonisins and deoxynivalenol in corn-based food products in Portugal
Martins HM, et al.
Food And Chemical Toxicology, 46(7), 2585-2587 (2008)
Selected mycotoxins affecting animal and human health
Handbook of Toxicologic Pathology, 699 (2002)
Determination of fumonisin B1 and B2 in corn and corn-based products in Turkey by high-performance liquid chromatography
OMURTAG GZ
Journal of Food Protection, 64, 1072-1075 (2001)
Zearalenone, deoxynivalenol and fumonisins in mixed-feed for laying hens
Martins HM, et al.
Mycotoxin Research, 22(4), 206-210 (2006)
Sonia Lo Magro et al.
Journal of food science, 76(1), T1-T4 (2011-05-04)
The occurrence of the fumonisins B(1) and B(2) in maize-based food products marketed in Italy was examined. A simply and reliable chromatographic method with fluorimetric detection and postcolumn o-phtalaldehyde derivatization was used for a monitoring of 100 samples (8 flours

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service