Skip to Content
Merck
All Photos(1)

Key Documents

24897

Supelco

Chloride ionophore I

Selectophore

Synonym(s):

meso-Tetraphenylporphyrin manganese(III)-chloride complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H28ClMnN4
CAS Number:
Molecular Weight:
703.11
Beilstein:
5699425
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

SMILES string

Cl[Mn]1n2c3ccc2c(-c4ccccc4)c5ccc(n5)c(-c6ccccc6)c7ccc(c(-c8ccccc8)c9ccc(n9)c3-c%10ccccc%10)n17

InChI

1S/C44H28N4.ClH.Mn/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;;/h1-28H;1H;/q-2;;+3/p-1/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;

InChI key

MIUMWNDEIWVIEG-YKKPBKTHSA-M

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Tetraphenylporphyrin manganese (III) chloride complex generally finds application in determining histidine in aqueous solutions by resonance light scattering technique.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chloride ionophore used in a cellulose acetate membrane electrode

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel histidine assay using tetraphenylporphyrin manganese (III) chloride as a molecular recognition probe by resonance light scattering technique.
Chen, Zhanguang, et al.
Analytica Chimica Acta, 570, 109-115 (2006)
Majid Moghadam et al.
Bioorganic & medicinal chemistry letters, 16(7), 2026-2030 (2006-01-24)
Mild and efficient oxidation of Hantzsch 1,4-dihydropyridines with sodium periodate catalyzed by Mn(TTP)Cl supported on polystyrene-bound imidazole is reported. This heterogeneous catalyst is of great stability and reusability in the oxidation of 1,4-dihydropyridines with sodium periodate without significant loss of
B Cheng et al.
Acta crystallographica. Section C, Crystal structure communications, 52 ( Pt 2), 361-363 (1996-02-15)
The crystal structure of [Mn(TPP)Cl] in space group I4/m (where TPP is C44H28N4) has been determined. The unit cell contains two full molecules, with one eighth of a molecule unique. An out-of-plane model for the Mn atom was applied and
M.J. Cha et al.
Analytica Chimica Acta, 315, 311-311 (1995)
Susan C Andrew et al.
Microorganisms, 9(1) (2021-01-21)
Prokaryotic communities coordinate quorum behaviour in response to external stimuli to control fundamental processes including inter-bacterial communication. The obligate intracellular bacterial pathogen Chlamydia adopts two developmental forms, invasive elementary bodies (EBs) and replicative reticulate bodies (RBs), which reside within a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service