Skip to Content
Merck

87252

Tetradodecylammonium nitrate

Selectophore, ≥99.0%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[CH3(CH2)10CH2]4N(NO3)
CAS Number:
Molecular Weight:
753.32
NACRES:
NB.61
PubChem Substance ID:
UNSPSC Code:
26111700
EC Number:
264-548-4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

for ion-selective electrodes

Quality Segment

product line

Selectophore

assay

≥99.0% (NT), ≥99.0%

form

flakes

mp

103-105 °C

SMILES string

[O-][N+]([O-])=O.CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C48H100N.NO3/c1-5-9-13-17-21-25-29-33-37-41-45-49(46-42-38-34-30-26-22-18-14-10-6-2,47-43-39-35-31-27-23-19-15-11-7-3)48-44-40-36-32-28-24-20-16-12-8-4;2-1(3)4/h5-48H2,1-4H3;/q+1;-1

InChI key

AHQMUYHFUYRUJV-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.
Tetradodecylammonium nitrate (TDDAN) is a quaternary ammonium salt, mainly used as electrolyte.

Application

Selected application examples
Ion-pair suitable for a nitrate-selective field-effect transistor membrane.
TDDAN has been used as solvent in the preparation of microelectrodes based on AgI-Ag2O-V2O5 glass transducer.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ion-pair suitable for a nitrate-selective field-effect transistor membrane

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany


pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library