Skip to Content
Merck
All Photos(2)

Documents

93206

Supelco

Carbonate ionophore VII

Selectophore, function tested

Synonym(s):

N,N-Dioctyl-3α,12α-bis(4-trifluoroacetylbenzoyloxy)-5β-cholan-24-amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H79F6NO7
CAS Number:
Molecular Weight:
1016.24
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

solid

quality

function tested

storage temp.

2-8°C

SMILES string

CCCCCCCCN(CCCCCCCC)C(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@@H](CC[C@]4(C)[C@H]3C[C@H](OC(=O)c5ccc(cc5)C(=O)C(F)(F)F)[C@]12C)OC(=O)c6ccc(cc6)C(=O)C(F)(F)F

InChI

1S/C58H79F6NO7/c1-6-8-10-12-14-16-34-65(35-17-15-13-11-9-7-2)50(66)31-18-38(3)46-29-30-47-45-28-27-43-36-44(71-53(69)41-23-19-39(20-24-41)51(67)57(59,60)61)32-33-55(43,4)48(45)37-49(56(46,47)5)72-54(70)42-25-21-40(22-26-42)52(68)58(62,63)64/h19-26,38,43-49H,6-18,27-37H2,1-5H3/t38-,43-,44-,45+,46-,47+,48+,49+,55+,56-/m1/s1

InChI key

PNOQSKXIBWVYCX-PNZPGYLPSA-N

General description

Carbonate ionophore VII is an ionophore mostly used in the preparation of ion selective electrodes.
Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Suk Choi et al.
Analytical chemistry, 74(10), 2435-2440 (2002-06-01)
Potentiometric properties of the PVC membrane-based electrodes prepared with molecular tweezer-type neutral carriers, 3,12-bis(TFAB)CA and deoxy-3,12-bis(TFAB)CA, and trifluoroacetyl-p-decylbenzene (TFADB) were measured in buffered electrolytes (0.1 M Tris-H2SO4, pH 8.6 and 8.0) and artificial seawater. It was observed that the deoxy-3,12-bis(TFAB)CA-based
Lee et al.
Analytical chemistry, 72(19), 4694-4699 (2000-10-12)
Potentiometric properties of the ion-selective electrodes) based on highly plasticized PVC membranes doped with the carbonate-selective cholic acid (CA) derivatives have been measured. The carbonate-selective neutral carriers have been prepared by coupling one to three trifluoroacetobenzoyl (TFAB) groups to a
A microsensor for carbonate ions suitable for microprofiling in freshwater and saline environments
De Beer, D., et al.
Limnology and Oceanography: Methods, 6, 532-541 (2008)
Potentiometric sensing array for monitoring aquatic systems.
Pankratova, Nadezda, et al.
Environmental Science. Processes & Impacts, 17.5, 906-914 (2015)
Mellisa T R Chikukwa et al.
Pharmaceutical development and technology, 25(3), 281-289 (2019-11-05)
The objective of the study was to mask the unpleasant taste of captopril (CPT). Taste masking was achieved by complexation of CPT with a basic ion exchange resin, Dowex® 66, using the batch method. Dowex® 66 was used for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service