Skip to Content
Merck

NA1111

GenElute Gel Extraction Kit

sufficient for 70 purifications

Synonym(s):

gel purification kit, Rapid purification of linear DNA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.52
UNSPSC Code:
41105501
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 70 purifications

Quality Segment

technique(s)

DNA purification: suitable

input

sample(s) (gel slices)

test parameters

sample volume: 3.5 g agarose

storage temp.

15-25°C

General description

The GenElute Gel Extraction Kit is designed for the rapid purification of linear and plasmid DNA fragments from standard or low-melting agarose gels. This kit can also be used to purify DNA from polyacrylamide gels. Typical recovery of DNA is up to 80%. Each column can bind up to 10 μg of DNA from up to a 3.5 g agarose slice.

Application

The isolated DNA is suitable for a variety of downstream applications, such as sequencing, PCR, restriction digestion, cloning and labeling.
GenElute Gel Extraction Kit has been used for the purification of DNA fragments from agarose gels.

Biochem/physiol Actions

The GenElute Gel Extraction Kit combines silica-binding technology with the convenience of a spin or vacuum column format. DNA fragments of interest are extracted from slices of an agarose gel and are bound to a silica membrane. Contaminants are removed by a simple spin or vacuum wash. The bound DNA is then eluted.

The purified DNA is suitable for a variety of downstream applications, such as automated DNA sequencing, PCR, restriction digestion, cloning, and labeling.
The GenElute Gel Extraction Kit combines silica-binding technology with the convenience of a spin or vacuum column format. DNA fragments of interest are extracted by solubilizing slices of an agarose gel. The Gel Solubilization Solution dissolves an agarose gel slice from gels run in TBE or TAE buffer. The extracted DNA fragments selectively adsorb onto the silica membrane in the presence of the Gel Solubilization Solution. Contaminants are removed by a simple spin or vacuum wash. Finally, the bound DNA is eluted in Tris buffer.

Features and Benefits

  • Bind up to 10 μg of DNA
  • Recoveries up to 80%
  • Up to 3.5 g can be processed per column
  • Compatible with both standard and low-melting agarose in TAE or TBE buffer

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC


Kit Components Only

Product No.
Description

  • Gel Solubilization Solution

  • Elution Solution (10 mM Tris-HCl, pH 9.0)

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Column Preparation Solution
    SDS

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1C - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library