Skip to Content
Merck
All Photos(1)

Key Documents

11126

Sigma-Aldrich

Potassium antimonyl tartrate trihydrate

puriss., meets analytical specification of USP, 99.0-103.0%, powder

Synonym(s):

Antimony potassium tartrate trihydrate, Tartar emetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4K2O12Sb2 · 3H2O
CAS Number:
Molecular Weight:
667.87
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
form:
powder

grade

puriss.

Quality Level

Assay

99.0-103.0%

form

powder

quality

meets analytical specification of USP

impurities

≤0.02 mmol/g free acid
≤0.02 mmol/g free alkali

loss

≤2.7% loss on drying, 105 °C

mp

≥300 °C (lit.)

cation traces

As: ≤150 mg/kg
Pb: ≤20 mg/kg

SMILES string

O.O.O.[K+].[K+].O=C1O[Sb-]23OC1C4O[Sb-]5(OC(C(O2)C(=O)O3)C(=O)O5)OC4=O

InChI

1S/2C4H4O6.2K.3H2O.2Sb/c2*5-1(3(7)8)2(6)4(9)10;;;;;;;/h2*1-2H,(H,7,8)(H,9,10);;;3*1H2;;/q2*-2;2*+1;;;;2*+3/p-4

InChI key

WBTCZEPSIIFINA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Potassium antimonyl tartrate trihydrate is a trivalent antimonial compound. It has been reported to be more toxic than pentavalent antimony (Pentostam) against Leishmania species. Its crystal structure has been investigated by the three-dimensional X-ray and white radiation neutron diffraction techniques. It crystallizes in space group C2221. Unit cell dimensions reported were: a = 11.192(2), b = 11.696(3), and c = 25.932(5)Å.

Application

Potassium antimonyl tartrate trihydrate was employed as an Sb(III)-containing drug to investigate its ability to induce cell death associated with DNA fragmentation in axenic amastigotes of Leishmania infantum
It may also be used as a standard solution to determine the metalloid concentration such as Sb(III) and Sb(V) in surface soil, using high performance liquid chromatography coupled to inductively coupled plasma-mass spectrometry (HPLC-ICP-MS).

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"A study of antimony complexed to soil-derived humic acids and inorganic antimony species along a Massachusetts highway"
Ceriotti G and Amarasiriwardena D
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 91(01), 85-93 (2009)
D Sereno et al.
Antimicrobial agents and chemotherapy, 45(7), 2064-2069 (2001-06-16)
The basic treatment of leishmaniasis consists in the administration of pentavalent antimonials. The mechanisms that contribute to pentavalent antimonial toxicity against the intracellular stage of the parasite (i.e., amastigote) are still unknown. In this study, the combined use of several
D Sereno et al.
Antimicrobial agents and chemotherapy, 41(5), 972-976 (1997-05-01)
Using a 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide microassay, previously described as a means of quantifying Leishmania amazonensis in vitro at the amastigote stage (D. Sereno and J. L. Lemesre, Parisitol. Res., in press), we have compared the activities of seven drugs, including those
Daniel B Liarte et al.
Parasitology research, 107(1), 205-212 (2010-04-08)
In the present study, we selected in vitro populations of Leishmania Viannia guyanensis, L.V. braziliensis, L. Leishmania amazonensis and L.L. infantum chagasi that were resistant to potassium antimony tartrate (SbIII). The resistance index of these populations varied from 4- to
Mostafa K Mohamed et al.
Journal of medical virology, 78(9), 1185-1189 (2006-07-19)
The origin of the hepatitis C virus (HCV) epidemic in Egypt has been attributed to intravenous schistosomiasis treatment in rural areas in the 1960s to 70s. The objective of this study was to estimate the HCV-related morbidity in a rural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service