Skip to Content
Merck

66720

Methylene Blue hydrate

purum p.a., suitable for microscopy, ≥97.0% (calc. based on dry substance, AT)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H18ClN3S · xH2O
CAS Number:
Molecular Weight:
319.85 (anhydrous basis)
UNSPSC Code:
12171500
NACRES:
NA.32
PubChem Substance ID:
EC Number:
200-515-2
Beilstein/REAXYS Number:
3599847
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum p.a.

Quality Segment

assay

≥97.0% (calc. based on dry substance, AT)

form

powder

mol wt

319.85 g/mol (anhydrous basis)

concentration

≤100% (Methylthioninium chloride hydrate)

color

green to dark green

solubility

ethanol: 0.01 g/10 mL, blue to very deep blue

λmax

656-666 nm in water

suitability

suitable for microscopy

storage temp.

room temp

SMILES string

O.[Cl-].CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C

InChI

1S/C16H18N3S.ClH.H2O/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;;/h5-10H,1-4H3;1H;1H2/q+1;;/p-1

InChI key

WQVSELLRAGBDLX-UHFFFAOYSA-M

General description

Methylene Blue (MB) is a basic histological dye.

Application

Methylene Blue hydrate has been used:
  • in the preparation of methylene blue (MB) solution to stain zymograms
  • to label the injection site in the tissue slices
  • to pre-treat mice via oral gavage to study its effects on noise-induced hearing loss

Biochem/physiol Actions

Methylene blue (MB) plays a key role in microbiology and pharmacology. It is mainly used in staining live organisms and in methemoglobinemia treatment. MB can be considered as a possible and safe vasoconstrictor. It has several applications in textile, paper and hair coloring sectors.

Other Notes

On the efficiency of methylene blue versus persulfate catalysis of polyacrylamide gels, as investigated by capillary zone electrophoresis
may contain up to 22% water


Still not finding the right product?

Explore all of our products under Methylene Blue hydrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library