Skip to Content
Merck
All Photos(1)

Key Documents

XX0060

Supelco

Xylenes

Histology

Synonym(s):

Dimethylbenzenes, Dimethylbenzene, Xylene mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12352005
EC Index Number:
215-535-7
application(s):
histology
bp:
137-140 °C (lit.)
vapor pressure:
18 mmHg ( 37.7 °C)

vapor pressure

18 mmHg ( 37.7 °C)

Quality Level

form

liquid

expl. lim.

7 %

impurities

≤0.05% Water (H2O)

evapn. residue

≤0.002%

color

APHA: ≤10

refractive index

n20/D 1.497 (lit.)

bp

137-140 °C (lit.)

transition temp

flash point 29.4 °C

density

0.86 g/mL at 25 °C (lit.)
~0.86-0.88 g/cm3 at 25 °C

application(s)

histology

shipped in

ambient

storage temp.

room temp

SMILES string

Cc1ccc(C)cc1.Cc2cccc(C)c2.Cc3ccccc3C

InChI

1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3

InChI key

MVZVDAGWAAZJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Clearing agent for routine histology and cytology applications.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Color (APHA): 10 max
Evaporation residue: 0.002% max
Form: Liquid
Water (H2O): 0.05% max

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service