Skip to Content
Merck
All Photos(2)

Key Documents

534056

Sigma-Aldrich

Xylenes

histological grade

Synonym(s):

Xylene mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21
grade:
histological grade
Assay:
≥75.0% (xylenes isomers)
application(s):
histology
bp:
137-140 °C (lit.)
vapor pressure:
18 mmHg ( 37.7 °C)

grade

histological grade

Quality Level

vapor density

3.7 (vs air)

vapor pressure

18 mmHg ( 37.7 °C)

Assay

≥75.0% (xylenes isomers)

form

liquid

expl. lim.

7 %

impurities

≤0.05% (water)
≤25% ethylbenzene

evapn. residue

<0.002%

refractive index

n20/D 1.497 (lit.)

bp

137-140 °C (lit.)

mp

-34 °C

density

0.86 g/mL at 25 °C (lit.)

application(s)

histology

storage temp.

room temp

SMILES string

Cc1ccc(C)cc1.Cc2cccc(C)c2.Cc3ccccc3C

InChI

1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3

InChI key

MVZVDAGWAAZJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xylenes are an isomeric mixture of aromatic hydrocarbons namely, o-xylene, m-xylene, and p-xylene. It is used as a solvent in the pharmaceutical, printing, rubber, cleaning agent, paint thinner, varnish, synthetic fiber, and leather industries.

Application

Xylenes (histological grade) can be used as a clearing agent in routine histology and cytology applications to remove paraffin from tissue section slides before staining.
It can be used in:      
  • Cresyl violet staining.     
  • Fluoro-Jade B staining process.      
  • Giemsa cell staining protocols.

Packaging

20 L in steel drum
500 mL in glass bottle
534056-4L , 4×4 L in glass bottle / 534056-4L-D in poly bottle

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron ionization of o-xylene, m-xylene and p-xylene.
Jiao CQ and Adams SF.
Chemical Physics Letters, 573, 24-28 (2013)
Experimental and modeling study of the oxidation of xylenes.
Battin-Leclerc F, et al.
International Journal of Chemical Kinetics, 38(4), 284-302 (2006)
Assessment of paraffin removal from prostate FFPE sections using transmission mode FTIR-FPA imaging.
Hughes C, et al.
Analytical Methods : Advancing Methods and Applications, 6(4), 1028-1035 (2014)
Fluoro-Jade B Staining for Neuronal Cell Death.
Laflamme N, et al.
Bio-protocol, 6(1) (2016)
A survey of household products for volatile organic compounds.
Sack TM, et al.
Atmos. Environ., Part A, 26(6), 1063-1070 (1992)

Related Content

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service