Skip to Content
Merck

1.08297

Xylene (isomeric mixture)

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Mixed Xylenes Solvent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.21
UNSPSC Code:
12191501
Grade:
ACS reagent
Bp:
136-145 °C/1013 hPa
Vapor pressure:
8 hPa ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. ISO, reag. Ph. Eur.

vapor pressure

8 hPa ( 20 °C)

product line

EMSURE®

form

liquid

dilution

(for analytical testing)

kinematic viscosity

<0.9 cSt(20 °C)

bp

136-145 °C/1013 hPa

mp

-25 °C

transition temp

flash point 24 °C

solubility

0.175 g/L

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

General description

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Application

Xylene (isomeric mixture) can be used:
  • as a solvent in various analytical techniques, including chromatography and spectroscopy.
  • in the analysis of environmental samples to detect and quantify xylene and its isomers in air, water, and soil.
  • as a solvent for the extraction of organic compounds from complex matrices.

Other Notes

Discover EMSURE - to be more than sure!
Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

75.2 °F

flash_point_c

24 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library