Recommended Products
form
liquid
Quality Level
packaging
pkg of 500 mL
manufacturer/tradename
Montage®
technique(s)
molecular cloning: suitable (BAC prep)
molecular cloning: suitable (plasmid miniprep)
shipped in
ambient
1 of 4
This Item | L1912 | L3289 | NA9604 |
---|---|---|---|
packaging pkg of 500 mL | packaging - | packaging - | packaging - |
manufacturer/tradename Montage® | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename - |
technique(s) molecular cloning: suitable (BAC prep) | technique(s) - | technique(s) - | technique(s) DNA purification: suitable |
shipped in ambient | shipped in - | shipped in - | shipped in - |
Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
General description
Application
All
Storage and Stability
Legal Information
Disclaimer
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
We manufacture IP and antibody purification agarose beads that purify immunoglobulins and IgG fractions. Our kits also provide a total antibody purification process
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service