Skip to Content
Merck
All Photos(1)

Key Documents

8.14536

Sigma-Aldrich

Methyl propyl ketone

for synthesis

Synonym(s):

Methyl propyl ketone, 2-Pentanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(O)CH2CH2CH3
CAS Number:
Molecular Weight:
86.13
MDL number:
UNSPSC Code:
12352115
EC Index Number:
203-528-1
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
101 °C/1013 hPa

grade

for synthesis
synthesis grade

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

449 °C

potency

3730 mg/kg LD50, oral (Rat)
6480 mg/kg LD50, skin (Rabbit)

expl. lim.

1.56-8.70 % (v/v)

dilution

(for synthesis)

bp

101 °C/1013 hPa

mp

-78 °C

transition temp

flash point 8 °C

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(CCC)C

InChI

1S/C5H10O/c1-3-4-5(2)6/h3-4H2,1-2H3

InChI key

XNLICIUVMPYHGG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 90.0 % (a/a)
Isobuthyl methyl ketone (GC, area%): ≤ 10.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.805 - 0.807
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F

Flash Point(C)

8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service