Skip to Content
Merck
All Photos(1)

Key Documents

650279

Sigma-Aldrich

Trypsin, Porcine, MS Grade

Synonym(s):

Porcine Trypsin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.54
Pricing and availability is not currently available.

grade

Proteomics Grade

Quality Level

form

lyophilized

specific activity

≥10,000 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

suitability

suitable for mass spectrometry

foreign activity

Chymostrypsin <3 BTEE units/mg solid

shipped in

wet ice

storage temp.

−20°C

General description

A chemically modified trypsin that is resistant to autolysis and degradation. Trypsin specifically hydrolyzes amides, peptides and proteins at the C-terminus side of arginine and lysine residues. This mass spectrometry grade trypsin is methylated, TPCK treated, and affinity purified. It is specifically designed for digestion of proteins for sequencing and peptide fragment analysis by mass spectrometry. A trypsin to protein ratio of 1:100 to 1:20 is recommended for mass spectrometry protocols. Note: 1 SET = 5 X 20 µg of modified trypsin, and 1 ml of resuspension buffer.

Warning

Toxicity: Harmful (C)

Unit Definition

One BAEE unit is defined as the ammount of enzyme required to produce a ΔA253 of 0.001 per min in a reaction volume of 3.2 ml at pH 7.6 at 25°C.
One BTEE unit is defined as the amount of enzyme required to hydrolyze 1 µmol BTEE per min at 25°C, pH 7.8

Reconstitution

Following reconstitution, aliquot and freeze (-°C). Unused stock solutions are stable for up to 2 weeks at -70°C. Avoid freeze/thaw cycles of solution.

Other Notes

Henzel, W.J., et al. 1993. PNAS90, 5011.
Rice, R.H., et al. 1977. Biochim Biophys Acta492, 316..

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service