Skip to Content
Merck
All Photos(1)

Key Documents

5.04594

Sigma-Aldrich

Gemcitabine, HCl

Synonym(s):

Gemcitabine, HCl, 2ʹ,2ʹ-Difluoro-2ʹ-deoxycytidine, HCl, 2ʹ-Deoxy-2ʹ,2ʹ-difluorocytidine, HCl, 4-Amino-1-((2R,5S)-3,3-difluoro-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, HCl, LY188011, dFdC, dFdCyd, Ribonucleotide Reductase Inhibitor II, RNR Inhibitor II, LY188011, dFdC, dFdCyd, 2ʹ,2ʹ-Difluoro-2ʹ-deoxycytidine, HCl, 2ʹ-Deoxy-2ʹ,2ʹ-difluorocytidine, HCl, 4-Amino-1-((2R,5S)-3,3-difluoro-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, HCl, Ribonucleotide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11F2N3O4 · xHCl
CAS Number:
Molecular Weight:
263.20 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 25 mg/mL

storage temp.

2-8°C

SMILES string

FC1([C@@H](O[C@@H]([C@H]1O)CO)[n]2[c](nc(cc2)N)=O)F.Cl

InChI

1S/C9H11F2N3O4.ClH/c10-9(11)6(16)4(3-15)18-7(9)14-2-1-5(12)13-8(14)17;/h1-2,4,6-7,15-16H,3H2,(H2,12,13,17);1H/t4-,6-,7-;/m1./s1

InChI key

OKKDEIYWILRZIA-OSZBKLCCSA-N

General description

A bioavailable, moderately toxic, deoxyctytidine analog with anti-metabolite and anti-tumor properties. Shown to be effective against a variety of tumors and blocks the growth of cancer cells in culture (IC 50 = 8.7, 57, 3.7, and 36 nM for L1210, CEM, Caco2, and HeLa cells, respectively). Following its uptake into cells by nucleoside transporters it is phosphorylated to its mono (dFdCMP), di (dFdCDP), and triphosphorylated (dFdCTP) forms by deoxycytidine kinase. dFdCDP and dFdCTP are reported to inhibit the activity of ribonucleotide reductase and impede DNA synthesis and repair mechanisms and induce cell death. Blocks the progression of cells through the G1/S phase. The dFdCTP is shown to compete with endogenous deoxycytidine triphosphate (dCTP) for incorporation into DNA and against CTP into RNA. Gemcitabine is also shown to block mitochondrial DNA polymerase γ. Works synergistically with other chemotherapeutic agents to enhance their cytotoxicity. Its half life is reported to be 42 to 94 min (depending upon the age and sex of the individual).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DNA
Reversible: no
Secondary Target
DNA polymerase

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Hung, S.W., et al. 2012. Cancer Lett.320, 138.
Fowler, J.D., et al. 2008. J. Biol. Chem.283, 15339.
Mackey, J.R., et al. 1998. Cancer Res.58, 4349.
Eda, H., et al. 1998. Cancer Res.58, 1165.
Burris III, H.A., et al. 1997. J. Clin. Oncol.15, 2403.
Heinemann, V., et al. 1992. Cancer Res.52, 533.
Hertel, L.W., et al. 1990. Cancer Res.50, 4417.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H A Burris et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 15(6), 2403-2413 (1997-06-01)
Most patients with advanced pancreas cancer experience pain and must limit their daily activities because of tumor-related symptoms. To date, no treatment has had a significant impact on the disease. In early studies with gemcitabine, patients with pancreas cancer experienced
Sau Wai Hung et al.
Cancer letters, 320(2), 138-149 (2012-03-20)
Clinical refractoriness to nucleoside analogs (e.g., gemcitabine, capecitabine) is a major scientific problem and is one of the main reasons underlying the extremely poor prognostic state of pancreatic cancer. The drugs' effects are suboptimal partly due to cellular mechanisms limiting
J R Mackey et al.
Cancer research, 58(19), 4349-4357 (1998-10-10)
Gemcitabine (2',2'-difluorodeoxycytidine) is a novel pyrimidine nucleoside drug with clinical efficacy in several common epithelial cancers. We have proposed that gemcitabine requires nucleoside transporter (NT) proteins to permeate the plasma membrane and to exhibit pharmacological activity. In humans, there are
H Eda et al.
Cancer research, 58(6), 1165-1169 (1998-03-27)
We showed that the efficacy of the new 2'-deoxycytidine (2'-dCyd) analogue antimetabolite 2'-deoxy-2'-methylidenecytidine (DMDC) correlates well with tumor levels of cytidine (Cyd) deaminase in human cancer xenograft models. DMDC was highly effective in tumors with higher levels of Cyd deaminase
Jason D Fowler et al.
The Journal of biological chemistry, 283(22), 15339-15348 (2008-04-02)
Gemcitabine, 2'-deoxy-2', 2'-difluorocytidine (dFdC), is a drug approved for use against various solid tumors. Clinically, this moderately toxic nucleoside analog causes peripheral neuropathy, hematological dysfunction, and pulmonary toxicity in cancer patients. Although these side effects closely mimic symptoms of mitochondrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service