Skip to Content
Merck
All Photos(1)

Key Documents

126658

Sigma-Aldrich

Albumin, Human Serum, Fraction V, High Purity

Synonym(s):

Albumin, Human Serum, Fraction V, High Purity

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

description

Merck USA index - 14, 8468

Assay

≥95% (cellulose acetate electrophoresis)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

electrophoresis: suitable

pI 

4.7

solubility

aqueous buffer: 5 mg/mL
water: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

General description

Human serum albumin (HSA) is an unglycosylated, soluble, and globular monomeric protein. It is abundantly found in the plasma, with a multidomain comprising three homologous domains, called I, II, and III. Three of these domains have subdomains A and B with common structural motifs. The HSA gene is localized on the human chromosome 4q13.3.

Application

Albumin, human serum, fraction V, high purity has been used to determine the binding affinity of nanobody α human serum albumin (HSA) to HSA and injected to detect the signals of tandem binding of human interleukin (IL-21R), IL-21-αHSA, and HSA. It has also been used to study the effects of fetuin-A phosphorylation on calciprotein particles (CPP) uptake.

Biochem/physiol Actions

Human serum albumin (HSA) plays a major role as a carrier protein for fatty acids, thyroid hormones, and steroids. It is involved in stabilizing extracellular fluid volume. HSA is used to treat hemodialysis, acute liver failure, chronic liver disease. It serves as an excipient for vaccines and therapeutic protein drugs. HSA is implicated in biological applications, including ligand trapping, nano delivery of drugs, and fusion proteins.

Warning

Toxicity: Standard Handling (A)

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Alternatively, this product may be stored in the refrigerator (4°C) prior to reconstitution. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanaya Chatterjee et al.
PloS one, 7(5), e37468-e37468 (2012-06-01)
Virstatin is a small molecule that inhibits Vibrio cholerae virulence regulation, the causative agent for cholera. Here we report the interaction of virstatin with human serum albumin (HSA) using various biophysical methods. The drug binding was monitored using different isomeric
G O Evans et al.
Laboratory animals, 20(1), 27-31 (1986-01-01)
Widely differing results were obtained for urinary protein determinations in male rats using 2 test strips and a quantitative Coomassie Blue sodium dodecyl sulphate method. A comparison of the sensitivity of the protein methods with respect to rat albumin and
Hongchuan Liu et al.
International immunopharmacology, 101(Pt A), 108307-108307 (2021-11-05)
Interleukin-21 (IL-21) has exhibited anti-tumor activity in preclinical and clinical studies; however, its modest efficacy and short half-time has limited its therapeutic utility as a monotherapy. Therefore, we engineered a fusion protein (IL-21-αHSA) in which a nanobody targeting human serum
L S Schlesinger et al.
Infection and immunity, 62(1), 280-289 (1994-01-01)
We have previously determined that complement receptors on human mononuclear phagocytes and complement component C3 in nonimmune serum mediate phagocytosis of the intracellular bacterial pathogen Mycobacterium leprae, the agent of leprosy. We have also determined that C3 fixes selectively to
L S Schlesinger et al.
The Journal of experimental medicine, 174(5), 1031-1038 (1991-11-01)
Previous studies from this laboratory have demonstrated that Mycobacterium leprae, an obligate intracellular bacterial parasite, enters human mononuclear phagocytes via complement receptors on these host cells and bacterium-bound C3. The present study investigates the role of M. leprae surface molecules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service