Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium azide
Synonym(s):
Sodium azide
Linear Formula:
NaN
3
CAS Number:
26628-22-8
Molecular Weight:
65.01
EC Number:
247-852-1
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (7)
Biological Source
Color
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Purum (1)
Purum P.A. (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
71289
Sodium azide,
BioUltra, ≥99.5% (T)
Expand
View Pricing
1.06688
Sodium azide,
extra pure, Technipur
®
Expand
View Pricing
71290
Sodium azide,
purum p.a., ≥99.0% (T)
Expand
View Pricing
8.22335
Sodium azide,
for synthesis
Expand
View Pricing
S2002
Sodium azide,
ReagentPlus
®
, ≥99.5%
Expand
View Pricing
769320
Sodium azide,
≥99%, ultra dry
Expand
View Pricing
S8032
Sodium azide,
BioXtra
Expand
View Pricing