Skip to Content
Merck
All Photos(1)

Key Documents

1.05030

Sigma-Aldrich

Potassium fluoride

extra pure, Technipur®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KF
CAS Number:
Molecular Weight:
58.10
Beilstein:
3902818
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

vapor pressure

1.3 hPa ( 885 °C)

Quality Level

pH

8-9 (20 °C, 50 g/L in H2O, slurry)

bp

>1505 °C/1013 hPa

mp

858 °C (lit.)

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium fluoride (KF) occurs naturally as the mineral carobbiite. It is mainly used as a fluorinating agent in chemical industries to produce a wide-range of fluorinated compounds by selective fluorination reactions. It also finds application in the electronic industry as a metal surface treatment product. Upon post-deposition treatment of KF in CuInGaSe2 (CIGS) based thin-film solar cells, power conversion efficiency is enhanced. This surface modification has a direct influence on solar cell performance leading to an increase in open-circuit voltage, fill factor, and net charge carrier density of the cell.

KF can also be used as:
  • A fluxing agent in the field of metallurgy for metal-joining operations, including welding, soldering, and brazing.
  • An etching agent in the manufacture of some optical glasses.
  • An additive in the formation of single crystals to change the crystal structure, crystal size, and crystal morphology.
A versatile fluoride ion source used in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elucidation of the role of potassium fluoride in the chemical and physical nature of ZSM-5 zeolite
Ke JA & Wang I
Materials Chemistry and Physics, 68(1-3), 157-165 (2001)
Potassium fluoride postdeposition treatment with etching step on both Cu-rich and Cu-poor CuInSe2 thin film solar cells
Babbe F, et al.
Physical Reiview Materials (2018)
Efficiency enhancement of Cu(In,Ga)Se2 thin-film solar cells by a post-deposition treatment with potassium fluoride
Laemmle A, et al.
Physica Status Solidi, 7(9), 631-634 (2013)
Crystal growth and characterization of ?-glycine grown from potassium fluoride for photonic applications
Dillip GR, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 79(5), 1123-1127 (2011)
Structural properties and chemical flux reactivities of aqueous suspensions of mixtures of potassium tetrafluoroborate and potassium fluoride
Ross RA, et al.
Thermochimica Acta, 107, 321-329 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service