Skip to Content
Merck
All Photos(2)

Key Documents

449148

Sigma-Aldrich

Potassium fluoride

anhydrous, powder, ≥99.9% trace metals basis

Synonym(s):

Potassium monofluoride, Potassium monofluoride (KF)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KF
CAS Number:
Molecular Weight:
58.10
Beilstein:
3902818
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

≥99.9% trace metals basis

form

powder

impurities

≤1000.0 ppm Trace Metal Analysis

mp

858 °C (lit.)

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium fluoride is a white crystalline solid that is highly soluble in water. It is a strong base widely used in the field of catalysis, polymer synthesis, and semiconductor devices. KF has a relatively high melting point. It dissolves well in hydrofluoric acid. However, KF is insoluble in alcohol.

Application

A versatile fluoride ion source used in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides.
Potassium fluoride can be used:
  • To fabricate CIGS thin films solar cells. Efficiency increased by up to 21.7% after KF postdeposition treatment. This was due to the formation of a K-containing layer on the CIGS surface, which resulted in increased surface reactivity due to the formation of a bilayer structure in which a K—In—Se species covers the CIGSe compound.
  • To fabricate 2D vanadium carbide MXene for supercapacitor electrodes.
  • To synthesize sulfonyl fluoride through electrochemical oxidative coupling.
  • As an electrolyte additive for Li-ion batteries.
  • As a catalyst for ring-opening fluorination of cyclic thioethers.
  • To synthesize solid base catalysts, KF/Ca−Mg−Al hydrotalcite for the production of Biodiesel via transesterification reactions.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An aryne triggered ring-opening fluorination of cyclic thioethers with potassium fluoride
Rong Fan, et al.
Chemical Communications (Cambridge, England), 54, 7081-7084 (2018)
Indian Journal of Physics, 70B, 311-311 (1996)
Biodiesel Synthesis Catalyzed by the KF/Ca?Mg?Al Hydrotalcite Base Catalyst
Lijing Gao, et al
Energy and Fuels, 24, 646-651 (2010)
Macromolecules, 29, 8391-8391 (1996)
Eur. J. Solid State Inorg. Chem., 33, 809-809 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service