Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium fluoride
Synonym(s):
Potassium fluoride
Linear Formula:
KF
CAS Number:
7789-23-3
Molecular Weight:
58.10
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (7)
Supelco (1)
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
ACS reagent (1)
Anhydrous (1)
Puriss (1)
Puriss P.A. (1)
Purum (1)
Purum P.A. (1)
certified reference material (1)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
402931
Potassium fluoride,
ACS reagent, ≥99.0%
Expand
View Pricing
449148
Potassium fluoride,
anhydrous, powder, ≥99.9% trace metals basis
Expand
View Pricing
1.04994
Potassium fluoride,
for analysis EMSURE
®
ACS
Expand
View Pricing
229814
Potassium fluoride,
≥99.97% trace metals basis
Expand
View Pricing
60238
Potassium fluoride,
BioUltra, ≥99.5% (F)
Expand
View Pricing
60240
Potassium fluoride,
purum p.a., ≥99.0% (F)
Expand
View Pricing
60239
Potassium fluoride,
puriss. p.a., ≥99.0% (F)
Expand
View Pricing
1.05030
Potassium fluoride,
extra pure, Technipur
®
Expand
View Pricing
NIST2203
Potassium fluoride,
NIST
®
SRM
®
2203, standard for ion-selective electrodes
Expand
View Pricing