Skip to Content
MilliporeSigma

Skip To

V-032

Voriconazole solution

2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Check Cart for Availability
$141.00

About This Item

Empirical Formula (Hill Notation):
C16H14F3N5O
CAS Number:
Molecular Weight:
349.31
UNSPSC Code:
41116107
NACRES:
NA.24

$141.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

2.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Fc1c(ncnc1)[C@@H]([C@](O)(C[n]3ncnc3)c2c(cc(cc2)F)F)C

InChI

1S/C16H14F3N5O/c1-10(15-14(19)5-20-7-22-15)16(25,6-24-9-21-8-23-24)12-3-2-11(17)4-13(12)18/h2-5,7-10,25H,6H2,1H3/t10-,16+/m0/s1

InChI key

BCEHBSKCWLPMDN-MGPLVRAMSA-N

General description

Voriconazole, sold under the trade name Vfend®, is a triazole antifungal drug prescribed for treatment of serious, invasive fungal infections. This certified Snap-N-Spike® Solution is suitable as starting material for linearity standards, calibrators, or controls for therapeutic drug monitoring or clinical and diagnostic testing of voriconazole in patient blood, serum, or plasma samples by HPLC or LC-MS/MS.

Legal Information

Vfend is a registered trademark of Pfizer, Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P-103F-031V-036
grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

format

single component solution

format

single component solution

format

single component solution

format

single component solution

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

packaging

ampule of 1 mL

packaging

ampule of 1 mL

packaging

ampule of 1 mL

packaging

ampule of 1 mL

concentration

2.0 mg/mL in methanol

concentration

2.0 mg/mL in methanol

concentration

2.0 mg/mL in methanol

concentration

1.0 mg/mL in methanol


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters