Skip to Content
Merck
All Photos(1)

Key Documents

S-038

Supelco

Spice Cannabinoid Mix solution

100 μg/mL each component in acetonitrile, ampule of 1.0 mL, certified reference material, Cerilliant®

Synonym(s):

Cannabinoid Mix for Spice Testing

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

(±)-CP 47, 497
(±)-CP 47, 497 C8 Homologue
HU-211
JWH-200
JWH-250

packaging

ampule of 1.0 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland)

concentration

100 μg/mL each component in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

multi-component solution

storage temp.

−20°C

General description

A multi-component mix containing Spice cannabinoids JWH-250; JWH-200; HU-211; (±)-CP 47,497; and (±)-CP 47,497 C8 homologue. On March 1, 2011, the US Drug Enforcement Agency (DEA) used its emergency power to classify five synthetic cannabinoids in Schedule I, the most restrictive category under the Controlled Substances Act. The five synthetic cannabinoids under this one year ban include JWH-018, JWH-073, JWH-200, CP-47,497 and its C8 homologue.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This study demonstrates the impact of various mobile phase modifiers on the separation; formate modifiers outperform acetate in terms of MS signals (or sensitivity) and chromatographic resolution.

Chromatograms

application for HPLCapplication for HPLC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service