Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Segment
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
composition
Cannabichromenic acid (CBCA), 500 μg/mL , Cannabidivarinic acid (CBDVA), 500 μg/mL , CBDA (Cannabidiolic acid), 500 μg/mL , CBGA (Cannabigerolic acid), 500 μg/mL , Tetrahydrocannabivarinic acid (THCVA), 500 μg/mL , THCA-A, 500 μg/mL
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
500 μg/mL (1% DIPEA and 0.05% Ascorbic acid in Acetonitrile,each analyte)
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
cannabis testing
format
multi-component solution
storage temp.
-65 to -96°C
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| grade certified reference material | grade certified reference material | grade - | grade - |
| format multi-component solution | format multi-component solution | format - | format - |
| packaging ampule of 1 mL | packaging ampule of 1 mL | packaging - | packaging - |
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) - | technique(s) - | technique(s) - |
| feature Snap-N-Spike®/Snap-N-Shoot® | feature second source lots, Snap-N-Spike®/Snap-N-Shoot® | feature - | feature - |
| application(s) cannabis testing | application(s) - | application(s) pharmaceutical | application(s) - |
Still not finding the right product?
Explore all of our products under Cannabinoid Mixture (Acids) - 6 Component solution
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_c
6.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




