Skip to Content

SML3348

JWH-018

≥98% (HPLC)

Synonym(s):

1-Naphthalenyl(1-pentyl-1H-indol-3-yl)methanone, 3-(1-Naphthoyl)-1-pentylindole, AM 678, JWH 018

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H23NO
CAS Number:
Molecular Weight:
341.45
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule I; Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CN(CCCCC)C2=C1C=CC=C2)C3=CC=CC4=C3C=CC=C4

InChI

1S/C24H23NO/c1-2-3-8-16-25-17-22(20-13-6-7-15-23(20)25)24(26)21-14-9-11-18-10-4-5-12-19(18)21/h4-7,9-15,17H,2-3,8,16H2,1H3

InChI key

JDNLPKCAXICMBW-UHFFFAOYSA-N

Biochem/physiol Actions

JWH-018, a synthetic cannabinoid, is a potent and selective full agonist at the CB1 and CB2 cannabinoid receptors. JWH-018 mimic the effects of Δ9-tetrahydrocannabinol (THC) and is found in several versions of the herbal mixture “Spice.”
Potent and selective full agonist at the CB1 and CB2 cannabinoid receptors

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library