Skip to Content
Merck
All Photos(1)

Key Documents

L-901

Supelco

Lorazepam solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10Cl2N2O2
CAS Number:
Molecular Weight:
321.16
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC1N=C(c2ccccc2Cl)c3cc(Cl)ccc3NC1=O

InChI

1S/C15H10Cl2N2O2/c16-8-5-6-12-10(7-8)13(19-15(21)14(20)18-12)9-3-1-2-4-11(9)17/h1-7,15,21H,(H,18,20)

InChI key

DIWRORZWFLOCLC-UHFFFAOYSA-N

General description

A Certified Spiking Solution® suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from clinical toxicology and forensic analysis to urine drug testing. Lorazepam, also known by trade names Ativan® or Orfidal, is a benzodiazepine often prescribed for treatment of anxiety disorders.

Legal Information

Ativan is a registered trademark of American Home Product Corp.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason T McMullan et al.
Prehospital emergency care : official journal of the National Association of EMS Physicians and the National Association of State EMS Directors, 17(1), 1-7 (2012-11-15)
The choice of the optimal benzodiazepine to treat prehospital status epilepticus is unclear. Lorazepam is preferred in the emergency department, but concerns about nonrefrigerated storage limits emergency medical services (EMS) use. Midazolam is increasingly popular, but its heat stability is
Olugbenga A Akingbola et al.
American journal of critical care : an official publication, American Association of Critical-Care Nurses, 21(6), 456-459 (2012-11-03)
A 6-year-old girl displayed symptoms of serotonin syndrome after accidental ingestion of Vyvanse (lisdexamfetamine dimesylate). Dexmedetomidine was administered because of persistent neuromuscular hyperactivity and severe agitation despite initial therapy with benzodiazepines. Some children show a paradoxical reaction to benzodiazepines, and
Natalie L Jacobowski et al.
Journal of psychiatric practice, 19(1), 15-28 (2013-01-22)
Delirious mania is a severe but under-recognized neuropsychiatric syndrome characterized by the rapid onset of delirium, mania, and psychosis, not associated with a prior toxicity, physical illness, or mental disorder. Catatonia is often a prominent feature of the syndrome. While
David W Loring et al.
Epilepsy & behavior : E&B, 25(3), 329-333 (2012-10-30)
A double-blind, placebo-controlled, crossover design was employed to determine whether acute lorazepam (2 mg orally) cognitive side effects would emerge in a differential age-dependent fashion in 15 young (mean age=22 years) and 12 older (mean age=64 years) subjects. Acute use
Arun G Selvaraj et al.
Psychogeriatrics : the official journal of the Japanese Psychogeriatric Society, 12(3), 211-213 (2012-09-22)
Electroconvulsive therapy is safe and effective in the treatment of depression in older individuals. Minor cognitive side effects of electroconvulsive therapy include acute postictal confusion and reversible short-term memory deficits. However, interictal delirium is uncommon in absence of risk factors.

Protocols

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service