Skip to Content
Merck
All Photos(1)

Documents

L-025

Supelco

(−)-Levamisole hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2S
CAS Number:
Molecular Weight:
204.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

C1CN2C[C@@H](N=C2S1)c3ccccc3

InChI

1S/C11H12N2S/c1-2-4-9(5-3-1)10-8-13-6-7-14-11(13)12-10/h1-5,10H,6-8H2/t10-/m1/s1

InChI key

HLFSDGLLUJUHTE-SNVBAGLBSA-N

General description

Levamisole, an antihelminthic drug used to treat worm infestations in humans and animals, is an adulterant and cutting agent found in cocaine sold in the US and Canada. From 2008-2009, levamisole was detected in 69% of cocaine samples seized by the US Drug Enforcement Administration (DEA).

Application


  • Immunomodulatory research: The study explored the potential immunomodulatory effects of levamisole hydrochloride in humans and farm animals, highlighting its utility in enhancing immune responses and its therapeutic potential across a range of conditions (Gholami et al., 2023).

  • Anthelmintic research: A study developed a real-time detection method for the levamisole resistance variant in Haemonchus contortus, illustrating the role of levamisole hydrochloride in understanding and managing anthelmintic resistance (Antonopoulos et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M P Myklebust et al.
British journal of cancer, 107(10), 1684-1691 (2012-10-27)
The aim of this study was to investigate the value of the cyclin D1 isoforms D1a and D1b as prognostic factors and their relevance as predictors of response to adjuvant chemotherapy with 5-fluorouracil and levamisole (5-FU/LEV) in colorectal cancer (CRC).
Kelly R Magliocca et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 71(3), 487-492 (2013-01-10)
Systemic complications of levamisole-adulterated cocaine (LAC) use have recently been described. The objective of this review is to increase awareness of these manifestations among oral and maxillofacial surgeons. LAC exposure through inhalation, nasal insufflation, or injection can induce cutaneous vasculopathy
L E Ferreira et al.
Experimental parasitology, 134(3), 327-332 (2013-04-16)
Despite the overall progress of sheep farming in Brazil, infections with the gastrointestinal parasite Haemonchus contortus represent one the most important problems in sheep production, aggravated by the increasing resistance of nematodes to traditional anthelmintic drugs caused by inadequate sheep
Ana Mbokeleng Tsotetsi et al.
Tropical animal health and production, 45(3), 751-761 (2012-10-16)
The present study was conducted to determine the prevalence and distribution of gastrointestinal helminths, to detect the presence of anthelmintic resistance in livestock from small-scale farms and to determine the level of helminthosis awareness among small-scale farmers in Gauteng Province
Xingxin Li et al.
Annals of hematology, 92(9), 1239-1247 (2013-04-27)
The appropriate management of patients with moderate aplastic anemia (mAA) remains to be unclear and controversial. A cohort of 118 patients with mAA received a novel immunosuppressive strategy of cyclosporine alternately combined with levamisole (CSA and LMS regimen), which included

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service