Skip to Content
Merck

Skip To

1359302

USP

Levamisole hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(−)-Tetramisole hydrochloride, (S)-(−)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride, L(−)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, Levamisole hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C11H12N2S · HCl
CAS Number:
Molecular Weight:
240.75
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
4358988
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

levamisole

manufacturer/tradename

USP

mp

228-230 °C

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.C1CN2C[C@@H](N=C2S1)c3ccccc3

InChI

1S/C11H12N2S.ClH/c1-2-4-9(5-3-1)10-8-13-6-7-14-11(13)12-10;/h1-5,10H,6-8H2;1H/t10-;/m1./s1

InChI key

LAZPBGZRMVRFKY-HNCPQSOCSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levamisole hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Levamisole Hydrochloride Tablets

Biochem/physiol Actions

Shows both immunostimulant and immunosuppressant effects, depending on several controllable factors. Very effective in treatment of ascariasis (hookworm infestation). Useful in chemotherapy of colorectal cancers, possibly due to its stimulation of IL-1 production and direct activation of macrophages.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L038000031742L-025
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

Cerilliant®

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

analytical standard

grade

certified reference material

format

neat

format

neat

format

neat

format

single component solution

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

mp

228-230 °C

mp

228-230 °C

mp

228-230 °C

mp

-

API family

levamisole

API family

levamisole

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Levamisole hydrochloride


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters