Skip to Content
Merck
All Photos(1)

Documents

E-011

Supelco

(1S,2R)-(+)-Ephedrine hydrochloride

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(1S,2R)-(+)-α-(1-Methylaminoethyl)benzyl alcohol hydrochloride, (1S,2R)-(+)-2-Methylamino-1-phenyl-1-propanol hydrochloride, D-α-(1-Methylaminoethyl)benzyl alcohol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH · HCl
CAS Number:
Molecular Weight:
201.69
Beilstein:
6118541
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

218-220 °C (lit.)

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl[H].CN[C@H](C)[C@@H](O)c1ccccc1

InChI

1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10-;/m1./s1

InChI key

BALXUFOVQVENIU-GHXDPTCOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ephedrine is a methamphetamine analog commonly used as a stimulant and appetite suppressant. It is also found as an active constituent in herbs used for traditional Chinese medicine, such as má huáng. This certified reference standard can be used Snap-N-Spike® as a starting material in the preparation of calibrators or controls for clinical or forensic applications or Snap-N-Shoot® in quality control testing of dietary supplements containing ephedrine.

Application


  • Efficacy of Ceftiofur in Dairy Cow Metritis: A randomized controlled trial evaluated systemic ceftiofur for metritis therapy in dairy cows, demonstrating significant efficacy in curing metritis, which is crucial for improving reproductive outcomes and milk production. This highlights ceftiofur′s role in veterinary medicine for treating bacterial infections in livestock (Menta et al., 2024).

  • Pharmacodynamic Modeling: A study on the predictive modeling of phenotypic antimicrobial susceptibility based on beta-lactamase resistance genes provides insights into the mechanisms of action of ceftiofur hydrochloride, suggesting ways to enhance its clinical effectiveness against resistant bacterial strains (Rahman et al., 2024).



Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noel Kong et al.
Journal of separation science, 37(21), 3015-3023 (2014-08-22)
A sensitive hydrophilic interaction liquid chromatography coupled with tandem mass spectrometry method was developed and validated for the simultaneous detection and quantification of etilefrine and oxilofrine in equine blood plasma and urine. The method is highly sensitive and specific with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service