Skip to Content
Merck
All Photos(1)

Key Documents

M-129

Supelco

(1R,2S)-(-)-N--Methylephedrine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Ephedra

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO
CAS Number:
Molecular Weight:
179.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC(C(O)c1ccccc1)N(C)C

InChI

1S/C11H17NO/c1-9(12(2)3)11(13)10-7-5-4-6-8-10/h4-9,11,13H,1-3H3/t9-,11-/m0/s1

InChI key

FMCGSUUBYTWNDP-ONGXEEELSA-N

General description

Methylephedrine, a sympathomimetic amine and a bronchodilator, is a component of several over-the-counter cough medications such as "BRON." Methylephedrine occasionally used in athletics as a performance enhancing drug for its stimulant effect. This Certified Snap-N-Spike® Solution is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from sports testing and urine drug testing to forensic analysis and clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingguo Hou et al.
Electrophoresis, 28(9), 1352-1363 (2007-05-01)
In this work, simultaneous separation of eight stereoisomers of ephedrine and related compounds ((+/-)-ephedrine, (+/-)-pseudoephedrine, (+/-)-norephedrine and (+/-)-N-methylephedrine) was accomplished using a polymeric chiral surfactant, i.e. polysodium N-undecenoxycarbonyl-L-leucinate (poly-L-SUCL) by chiral (C)MEKC-ESI-MS. The conditions of CMEKC were first investigated. The
Satoshi Murao et al.
Internal medicine (Tokyo, Japan), 47(11), 1013-1015 (2008-06-04)
We report a 35-year-old man who was referred to our hospital with generalized convulsion and mixed acidosis presumably caused by abuse of SS-BRON tablets, an over-the-counter (OTC) antitussive medication sold in Japan. These tablets contain dihydrocodeine phosphate, methylephedrine, chlorpheniramine, and
Kathlyn A Parker et al.
Organic letters, 10(7), 1349-1352 (2008-03-05)
A short sequence based on asymmetric catalysis, chirality transfer, and an optimized carbometallation protocol gave an anti,anti stereotriad building block in six steps. Both enantiomers of the chirality source, N-methyl ephedrine, are inexpensive, and the auxiliary is recoverable. In one
Yan-Ming Liu et al.
Electrophoresis, 30(8), 1406-1411 (2009-03-19)
A new approach for the determination of methylephedrine hydrochloride (ME), thebaine, codeine phosphate (CP), and acetylcodeine (AC) was established by CE-ECL with ionic liquid (1-butyl-3-methylimidazolium tetrafluoroborate). The conditions for the CE separation, ECL detection, and the effect of ionic liquid
Yan-Ming Liu et al.
Biomedical chromatography : BMC, 23(11), 1138-1144 (2009-06-06)
A novel method for the determination of ephedra alkaloids (methylephedrine and pseudoephedrine) was developed by electrophoresis capillary (CE) separation and electrochemiluminesence detection (ECL). The use of ionic liquid (1-butyl-3-methylimidazolium tetrafluoroborate, BMIMBF(4)) improved the detection sensitivity markedly. The conditions for CE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service