Skip to Content
Merck
All Photos(4)

Key Documents

287636

Sigma-Aldrich

(1S,2S)-(+)-Pseudoephedrine

98%

Synonym(s):

(+)-Pseudoephedrine, (+)-ψ-Ephedrine, (1S,2S)-2-Methylamino-1-phenyl-1-propanol, d-Isoephedrine, d-Pseudoephedrine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO
CAS Number:
Molecular Weight:
165.23
Beilstein:
2414132
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]20/D +52°, c = 0.6 in ethanol

mp

118-120 °C

functional group

amine
hydroxyl
phenyl

SMILES string

CN[C@@H](C)[C@@H](O)c1ccccc1

InChI

1S/C10H15NO/c1-8(11-2)10(12)9-6-4-3-5-7-9/h3-8,10-12H,1-2H3/t8-,10+/m0/s1

InChI key

KWGRBVOPPLSCSI-WCBMZHEXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1S,2S)-(+)-Pseudoephedrine, a natural enantiomer of ephedrine, is a decongestant commonly used in cold and allergy medicines.

Application

(1S,2S)-(+)-Pseudoephedrine condenses with N,N-diisopropyl-2-formyl-1-naphthamide to form the corresponding oxazolidine derivative as a single diasterisomer.
(1S,2S)-(+)-Pseudoephedrine may be used as a chiral auxillary in asymmetric synthesis of enantioenriched organic compounds. It may also be used to prepare a novel tertiary pseudo C2-symmetric 1,2-diamine, which facilitates the enantioselective addition of methyl lithium to imines with better yield.

Biochem/physiol Actions

Non-selective adrenergic agonist; decongestant

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pseudoephenamine: a practical chiral auxiliary for asymmetric synthesis.
Marvin R Morales et al.
Angewandte Chemie (International ed. in English), 51(19), 4568-4571 (2012-03-31)
Wayland Hsiao et al.
BJU international, 110(8), 1196-1200 (2012-01-12)
What's known on the subject? and What does the study add? Modern surgical techniques have allowed preservation of fertility in most patients after post-chemotherapy retroperitoneal lymph node dissection (PC-RPLND), but some patients still have infertility after surgery. We reviewed our
James K Cunningham et al.
Drug and alcohol dependence, 126(1-2), 55-64 (2012-05-18)
Clandestine laboratory operators commonly extract ephedrine and pseudoephedrine-precursor chemicals used to synthesize methamphetamine-from over-the-counter cold/allergy/sinus products. To prevent this activity, two states, Oregon in 07/2006 and Mississippi in 07/2010, implemented regulations classifying ephedrine and pseudoephedrine as Schedule III substances, making
Jillian M Hagel et al.
Trends in plant science, 17(7), 404-412 (2012-04-17)
Amphetamine analogs are produced by plants in the genus Ephedra and by Catha edulis, and include the widely used decongestants and appetite suppressants pseudoephedrine and ephedrine. A combination of yeast (Candida utilis or Saccharomyces cerevisiae) fermentation and subsequent chemical modification
Beatriz Alonso et al.
The Journal of organic chemistry, 78(2), 614-627 (2012-12-25)
We have developed an efficient protocol for carrying out the stereocontrolled formal conjugate addition of hydroxycarbonyl anion equivalents to α,β-unsaturated carboxylic acid derivatives using (S,S)-(+)-pseudoephedrine as chiral auxiliary, making use of the synthetic equivalence between the heteroaryl moieties and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service