Skip to Content
Merck
All Photos(1)

Documents

857501P

Avanti

16:0-17:0 cyclo PC

Avanti Research - A Croda Brand 857501P, powder

Synonym(s):

1-palmitoyl-2-cis-9,10-methylenehexadecanoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H80NO8P
CAS Number:
Molecular Weight:
746.05
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857501P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857501P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCC1C(C1)CCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

General description

16:0-17:0 cyclo PC or 1-palmitoyl-2-cis-9,10-methylenehexadecanoyl-sn-glycero-3-phosphocholine is a cyclopropane modified analog of PC. It is a glycerophospholipid and is the most important constituent of the eukaryotic cell membrane. Generally, it constitutes up to 40-50% of the cells and organelles.

Biochem/physiol Actions

Phosphatidylcholine (PC) is crucial for the synthesis of second messengers and lipid mediators. It also acts as an intermediate for the synthesis of other glycerophospholipid classes along with phosphatidylethanolamine (PE). Any disturbance in PC levels leads to various metabolic disorders.

Packaging

5 mL Amber Glass Screw Cap Vial (857501P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selma Maric et al.
Applied microbiology and biotechnology, 99(1), 241-254 (2014-10-11)
Phosphatidylcholine (PC) is a major component of eukaryotic cell membranes and one of the most commonly used phospholipids for reconstitution of membrane proteins into carrier systems such as lipid vesicles, micelles and nanodiscs. Selectively deuterated versions of this lipid have
Lian Shan et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 864(1-2), 22-28 (2008-02-12)
Lysophosphatidic acid (LPA) is a class of lipids that play multiple biological functions. Several reports show that they are potential biomarkers for diagnosing ovarian cancer. Therefore, it is necessary to accurately quantify their levels in biological samples. Here we report
Nicholas C Zitomer et al.
The Journal of biological chemistry, 284(8), 4786-4795 (2008-12-20)
Fumonisin B(1) (FB(1)) is a mycotoxin that inhibits ceramide synthases (CerS) and causes kidney and liver toxicity and other disease. Inhibition of CerS by FB(1) increases sphinganine (Sa), Sa 1-phosphate, and a previously unidentified metabolite. Analysis of the latter by
Jelske N van der Veen et al.
Biochimica et biophysica acta. Biomembranes, 1859(9 Pt B), 1558-1572 (2017-04-16)
Phosphatidylcholine (PC) and phosphatidylethanolamine (PE) are the most abundant phospholipids in all mammalian cell membranes. In the 1950s, Eugene Kennedy and co-workers performed groundbreaking research that established the general outline of many of the pathways of phospholipid biosynthesis. In recent
Junliang Wan et al.
Journal of agricultural and food chemistry, 67(46), 12953-12961 (2019-10-23)
Most common sphingolipids are comprised of "typical" sphingoid bases (sphinganine, sphingosine, and structurally related compounds) and are produced via the condensation of l-serine with a fatty acyl-CoA by serine palmitoyltransferase. Some organisms, including mammals, also produce "atypical" sphingoid bases that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service