Skip to Content
Merck
All Photos(2)

Key Documents

857138C

Avanti

18:2 Lyso PA

Avanti Research - A Croda Brand

Synonym(s):

1-(9Z,12Z-octadecadienoyl)-2-hydroxy-sn-glycero-3-phosphate (sodium salt); PA(18:2(9Z,12Z)/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H38NaO7P
CAS Number:
Molecular Weight:
456.49
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1-linoleoyl-2-hydroxy-sn-glycero-3-phosphate (sodium salt), chloroform

Assay

>99% (LPA; may contain up to 10% of the 2-LPA isomer, TLC)

form

liquid

packaging

pkg of 1 × 1 mL (857138C-10mg)
pkg of 1 × 2.5 mL (857138C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (857138C-10mg)
10 mg/mL (857138C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Lysophosphatidic acid (LPA) is a lysophospholipid comprising of glycerol 3-phosphate (G3P), fatty acids, and phosphates.

Biochem/physiol Actions

18:2 Lysophosphatidic acid (LPA) is an active ingredient of gintonin, which is a bioactive ligand of ginseng. LPA is synthesized in plants by the activity of the enzyme glycerol 3-phosphate acyltransferase (GPAT). LPA is an intermediate in the synthesis of phosphatidic acid (PA) and other phospholipids. Abnormal LPA metabolism is found in children with autism.

Packaging

5 mL Clear Glass Sealed Ampule (857138C-10mg)
5 mL Clear Glass Sealed Ampule (857138C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potential serum biomarkers from a metabolomics study of autism
Wang H, et al.
Journal of Psychiatry & Neuroscience, 41(1), 27-27 (2016)
Bioactive lipids in gintonin-enriched fraction from ginseng
Cho HJ, et al.
Journal of ginseng research (2017)
Ross Dobie et al.
Cell reports, 29(7), 1832-1847 (2019-11-14)
Iterative liver injury results in progressive fibrosis disrupting hepatic architecture, regeneration potential, and liver function. Hepatic stellate cells (HSCs) are a major source of pathological matrix during fibrosis and are thought to be a functionally homogeneous population. Here, we use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service