Skip to Content
Merck
All Photos(2)

Key Documents

857328P

Avanti

18:1 Cyclic LPA

1-oleoyl-sn-glycero-2,3-cyclic-phosphate (ammonium salt), powder

Synonym(s):

1-(9Z-octadecenoyl)-sn-glycero-2,3-cyclic-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H42NO6P
CAS Number:
Molecular Weight:
435.54
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857328P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857328P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCCCCCC/C=C\CCCCCCCC)OC[C@@H]1COP(O1)([O-])=O.[NH4+]

InChI

1S/C21H39O6P.H3N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)25-18-20-19-26-28(23,24)27-20;/h9-10,20H,2-8,11-19H2,1H3,(H,23,24);1H3/b10-9-;/t20-;/m1./s1

InChI key

BQDHONSGIYYBPK-JGSYTFBMSA-N

General description

Cyclic phosphatidic acid (cPA) is a naturally occurring analog of the growth factor-like phospholipid mediator, lysophosphatidic acid (LPA). The sn-2 hydroxy group of CPA forms a 5-membered ring with the sn-3 phosphate. cPA affects numerous cellular functions, including anti-mitogenic regulation of the cell cycle, induction of stress fiber formation, inhibition of tumor cell invasion and metastasis, and regulation of differentiation and survival of neuronal cells.
Interestingly, many of these cellular responses caused by cPA oppose those of LPA despite the activation of apparently overlapping receptor populations.

Application

18:1 Cyclic LPA has been used for the preparation of liposomes. It has also been used for the extraction recovery of cyclic phosphatidic acid (cPA) at different pH conditions using Bligh−Dyer method.

Packaging

5 mL PTFE Vial with Screw Cap (857328P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of cyclic phosphatidic acid in human serum by LC/ESI/MS/MS
Shan L, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 862(1-2), 161-167 (2008)
Kitti Koprivanacz et al.
Cellular signalling, 32, 66-75 (2017-01-21)
Src homology 3 or SH3 domains constitute one of the most common protein domains in signal transduction, generally characterized by their binding to proline-rich sequences on interacting signaling proteins. Caskin1, a scaffold protein regulating cortical actin filaments, enriched in neural
Naturally occurring analogs of lysophosphatidic acid elicit different cellular responses through selective activation of multiple receptor subtypes.
Fischer DL, et al.
Molecular Pharmacology, 54, 979-988 (1998)
Inhibition of tumor invasion and metastasis by a novel lysophosphatidic acid (cyclic LPA).
Mukai M, et al.
International Journal of Cancer. Journal International Du Cancer, 81, 918-922 (1999)
Cyclic phosphatidic acid elicits neurotrophin-like actions in embryonic hippocampal neurons.
Fujiwara Y, et al.
Journal of Neurochemistry, 87, 1272-1283 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service