Skip to Content
Merck
All Photos(2)

Key Documents

846725P

Avanti

18:1 Lyso PE

Avanti Research - A Croda Brand

Synonym(s):

1--(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine; PE(18:1(9Z)/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H46NO7P
CAS Number:
Molecular Weight:
479.59
UNSPSC Code:
51191904
NACRES:
NA.25

description

1-oleoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine

Assay

>99% (LPE; may contain up to 10% of the 2-LPE isomer, TLC)

form

powder

packaging

pkg of 1 × 1 g (846725P-1g)
pkg of 2 × 100 mg (846725P-200mg)
pkg of 1 × 25 mg (846725P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[NH3+])=O)([H])COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C23H46NO7P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-23(26)29-20-22(25)21-31-32(27,28)30-19-18-24/h9-10,22,25H,2-8,11-21,24H2,1H3,(H,27,28)/b10-9-/t22-/m1/s1

InChI key

PYVRVRFVLRNJLY-MZMPXXGTSA-N

General description

Lysophosphatidyletnolamine (LPE), an enigmatic lipid of bacteria is obtained from phosphatidylethanolamine during various fluctuations of the bacterial growth conditions.

Application

18:1 Lyso PE is suitable for use in the synthesis of lipid molecules. It is also suitable to use as a standard in the study of systematic optimization of hydrophilic interaction liquid chromatography-mass spectrometry (HILIC/MS) separation of acidic lipid classes.

Packaging

5 mL Clear Glass Sealed Ampule (846725P-200mg)
5 mL Clear Glass Sealed Ampule (846725P-25mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (846725P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Opposite effects of lysophosphatidylethanolamines on conformation of OmpF-like porin from Yersinia pseudotuberculosis
Davydova AL, et al.
Protein and peptide letters, 22(12), 1060-1065 (2015)
Hydrophilic interaction liquid chromatography? mass spectrometry of (lyso) phosphatidic acids,(lyso) phosphatidylserines and other lipid classes
Cifkova E, et al.
Journal of Chromatography A, 1439, 65-73 (2016)
Enzymatic formation of N-acylethanolamines from N-acylethanolamine plasmalogen through N-acylphosphatidylethanolamine-hydrolyzing phospholipase D-dependent and-independent pathways
Tsuboi K, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1811(10), 565-577 (2011)
Svenja Bockelmann et al.
Journal of lipid research, 59(3), 515-530 (2018-01-19)
Ceramides are central intermediates of sphingolipid metabolism with dual roles as mediators of cellular stress signaling and mitochondrial apoptosis. How ceramides exert their cytotoxic effects is unclear and their poor solubility in water hampers a search for specific protein interaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service