Skip to Content

P7693

3-sn-Phosphatidylethanolamine from bovine brain

Type I, ≥98% (TLC), lyophilized powder

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphoethanolamine, L-α-Cephalin, L-α-Phosphatidylethanolamine from bovine brain

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
51191904
EC Number:
292-752-3
NACRES:
NA.77
Assay:
≥98% (TLC)
Form:
lyophilized powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


type

Type I

Quality Segment

assay

≥98% (TLC)

form

lyophilized powder

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OCC[N+H3])OCC(OC(=O)C)COC(=O)C

InChI

1S/C9H18NO8P/c1-7(11)15-5-9(18-8(2)12)6-17-19(13,14)16-4-3-10/h9H,3-6,10H2,1-2H3,(H,13,14)

InChI key

CFWRDBDJAOHXSH-UHFFFAOYSA-N

Biochem/physiol Actions

The major structural phospholipid in brain, comprising 20-25% of total lipid; primarily localized to gray matter.

Packaging

Packaged under Argon.

Disclaimer

Purity when prepared is 98-99%. Solutions exposed to room temperature can decompose 0.3-0.5% per day.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library