L4129
L-α-Lysophosphatidylcholine from egg yolk
≥99%, Type I, powder
Synonym(s):
1-Acyl-sn-glycero-3-phosphocholine, L-α-Lysolecithin
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Recommended Products
biological source
egg yolk
description
zwitterionic
type
Type I
Assay
≥99%
form
powder
color
white
functional group
phospholipid
lipid type
phosphoglycerides
shipped in
ambient
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
L-α-Lysophosphatidylcholine from egg yolk has been used:
- as a surfactant
- as a media supplement and in Staphylococcus aureus inhibition assay in dermatitis
- as a demyelinating agent for nerve
- as a component in sperm nuclei preparation buffer
Biochem/physiol Actions
Lysophosphatidylcholine is a major component of oxidized low density lipoproteins, and has been implicated in various inflammatory reactions, including atherosclerosis. It is used to demyelinate spinal neurons and study the processes underlying remyelination. It activates protein kinase C, p38 MAP kinase, p42 MAP Kinase, and the jun kinase (JNK) pathway, and stimulates transcription of c-jun. Lysophosphatidylcholine accumulates during cardiac ischemia and may induce arrhythmias by uncoupling gap junction communication, and increase ischemic damage by enhancing Na+ loading in cardiac myocytes. It also activates TREK1, TREK2 and TRAAK K+ channels.
Quality
Contains primarily palmitic and stearic acids.
Preparation Note
Prepared by the action of phospholipase A on egg L-α-phosphatidylcholine.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Scientific reports, 3, 1287-1287 (2013-02-16)
Genetic therapies for cystic fibrosis (CF) must be assessed for safety and efficacy, so testing in a non-human primate (NHP) model is invaluable. In this pilot study we determined if the conducting airways of marmosets (n = 2) could be
Frontiers in neuroscience, 11, 433-433 (2017-08-22)
In this work a new MRI modality entitled Relaxation Along a Fictitious Field in the rotating frame of rank 4 (RAFF4) was evaluated in its ability to detect lower myelin content in lysophosphatidyl choline (LPC)-induced demyelinating lesions. The lesions were
Transplantation of human skin microbiota in models of atopic dermatitis
JCI insight, 1(10), 031712-031712 (2016)
Director configurations in nematic droplets with inhomogeneous boundary conditions
Physical review. E, 72(3), 031712-031712 (2005)
PloS one, 8(9), e76233-e76233 (2013-12-07)
Lysophosphatidylcholine (LPC) is the main phospholipid component of oxidized low-density lipoprotein (oxLDL) and is usually noted as a marker of several human diseases, such as atherosclerosis, cancer and diabetes. Some studies suggest that oxLDL modulates Toll-like receptor (TLR) signaling. However
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service