Skip to Content
Merck
All Photos(2)

Key Documents

840028P

Avanti

17:0 PS

Avanti Research - A Croda Brand

Synonym(s):

PS(17:0/17:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H77NO10PNa
CAS Number:
Molecular Weight:
786.00
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-diheptadecanoyl-sn-glycero-3-phospho-L-serine (sodium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840028P-10mg)
pkg of 1 × 25 mg (840028P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C40H78NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-38(42)48-33-36(34-49-52(46,47)50-35-37(41)40(44)45)51-39(43)32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h36-37H,3-35,41H2,1-2H3,(H,44,45)(H,46,47);/q;+1/p-1/t36-,37+;/m1./s1

InChI key

HNPCQASYRKKJRQ-JBEIVTSCSA-M

General description

Phosphatidylserine (PtdSer) is an anionic phospholipid abundantly present in the eukaryotic cell. 10% of the total cellular lipid consists of phosphatidylserine. O‐phospho‐L‐serine is a heat‐stable substrate.

Application

17:0 PS has been used as an internal standard for BXD liver and plasma sample analysis. It may be used as a standard to estimate the cellular lipidome of Saccharomyces cerevisiae by liquid chromatography coupled with tandem mass spectrometry.

Biochem/physiol Actions

Phosphatidylserine (PtdSer) participates in various intracellular signaling pathways. It also play a major role in apoptosis and blood clotting.

Packaging

5 mL Amber Glass Screw Cap Vial (840028P-10mg)
5 mL Amber Glass Screw Cap Vial (840028P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karamat Mohammad et al.
Journal of visualized experiments : JoVE, (157) (2020-03-24)
Lipids are structurally diverse amphipathic molecules that are insoluble in water. Lipids are essential contributors to the organization and function of biological membranes, energy storage and production, cellular signaling, vesicular transport of proteins, organelle biogenesis, and regulated cell death. Because
Koshiki Mino et al.
FEBS letters, 551(1-3), 133-138 (2003-09-11)
O-Acetylserine sulfhydrylase (OASS), a pyridoxal 5'-phosphate (PLP)-dependent enzyme, catalyzes the synthesis of L-cysteine from O-acetyl-L-serine and sulfide. O-Acetyl-L-serine is labile at high temperatures at which hyperthermophilic archaea live. Herein, a study of the substrate specificity of OASS from Aeropyrum pernix
Jason G Kay et al.
Cell communication and signaling : CCS, 17(1), 126-126 (2019-10-17)
Phosphatidylserine (PtdSer), an essential constituent of eukaryotic membranes, is the most abundant anionic phospholipid in the eukaryotic cell accounting for up to 10% of the total cellular lipid. Much of what is known about PtdSer is the role exofacial PtdSer
Pooja Jha et al.
Cell systems, 6(6), 722-733 (2018-06-18)
The genetics of individual lipid species and their relevance in disease is largely unresolved. We profiled a subset of storage, signaling, membrane, and mitochondrial liver lipids across 385 mice from 47 strains of the BXD mouse population fed chow or
Emily L Gill et al.
Journal of proteome research, 19(1), 424-431 (2019-11-13)
Parkinson's disease (PD) is characterized by the loss of dopaminergic neurons in the substantia nigra pars compacta of the brain, as well as the degeneration of motor and nonmotor circuitries. The cause of neuronal death is currently unknown, although chronic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service