Skip to Content
Merck
All Photos(2)

Key Documents

850367P

Avanti

19:0 PC

Avanti Research - A Croda Brand

Synonym(s):

PC(19:0/19:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H92NO8P
CAS Number:
Molecular Weight:
818.20
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dinonadecanoyl-sn-glycero-3-phosphocholine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850367P-1g)
pkg of 1 × 200 mg (850367P-200mg)
pkg of 1 × 25 mg (850367P-25mg)
pkg of 1 × 500 mg (850367P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCCC)=O)=O

InChI key

CJXPNBSAXZBLEC-USYZEHPZSA-N

General description

Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It the most common phospholipid in mammalian membranes. It is also an important component of the mucosal layer of the colon.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

19:0 PC is suitable to use as a phospholipid standard to for fatty acid analysis in Sinorhizobium meliloti. It may also be suitable for use to study the thermodynamics of phospholipids.

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance. It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.

Packaging

20 mL Clear Glass Screw Cap Vial (850367P-1g)
20 mL Clear Glass Screw Cap Vial (850367P-500mg)
5 mL Amber Glass Screw Cap Vial (850367P-200mg)
5 mL Amber Glass Screw Cap Vial (850367P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of three GC/MS methodologies for the analysis of fatty acids in Sinorhizobium meliloti: Development of a micro-scale, one-vial method
Basconcillo LS and McCarry BE
Journal of Chromatography. B, Biomedical Sciences and Applications, 871(1), 22-31 (2008)
Integrative Medicine - E-Book (2017)
The Membranes of Cells (2016)
Thermodynamics of the gel to liquid crystal 1, 2-diacylphosphatidylcholines transition. High-pressure microcalorimetry
Potekhin SA, et al.
Thermochimica Acta, 560, 17-26 (2013)
Hsiu-Ching Hsu et al.
Journal of biomedical science, 21, 101-101 (2014-11-20)
Use of the chemotherapeutic drug doxorubicin (DOX) is associated with serious cardiotoxicity, as it increases levels of reactive oxygen species (ROS). N-3 polyunsaturated fatty acid dietary supplements can be of benefit to patients undergoing cancer therapy. The aims of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service