Z679046
Tamper evident sample tube
non-graduated, capacity 7 mL
About This Item
Recommended Products
material
clear tube
colorless tube
flat bottom
polyethylene cap
polypropylene
polypropylene tube
screw top cap
white
sterility
non-sterile
feature
graduations: no
neck thread EXTERNAL THREAD
packaging
pack of 1,000 ea
manufacturer/tradename
Simport T552-7ATTP
capacity
7 mL
tube diam.
13.4 mm
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
These unique transport tubes incorporate a seal that needs to be broken in order to open it, which leaves an obvious visual indication that the vial has been opened.
Manufactured from non-toxic polypropylene, these tubes provide strength and clarity and exhibit some unique design features. Five sizes are available from 5 to 30 mL. All graduated tubes are in 0.5 mL increments. They have external threads to provide a smooth and uniform inner surface.
Caps should be ordered separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service