About This Item
NACRES:
NB.17
UNSPSC Code:
41122800
Manufacturer/tradename:
Heathrow Scientific HS24312A
Feature:
to hold 96 tube(s) 10-13 mm tubes
Material:
white , fiberglass , polypropylene
Packaging:
pack of 2 ea
material
white , fiberglass , polypropylene
feature
to hold 96 tube(s) 10-13 mm tubes
packaging
pack of 2 ea
manufacturer/tradename
Heathrow Scientific HS24312A
L × W × H
82 mm × 126 mm × 69 mm
General description
The Peg Tube Rack is strong, tough, and versatile for bench or tube storage. It is constructed of fiberglass-reinforced polypropylene, which is ideal for bench work or tube storage.
Features and Benefits
Constructed of fiberglass-reinforced polypropylene
Spacious peg design allows easy viewing of tube contents
Use as a drying rack for glass plates or upside-down tubes
Pegs remain straight even after repeated autoclaving
Embossed alphanumeric identification for quick location and retrieval of tubes
Unique tear drop well design allows for better retention of tubes
Autoclavable
Spacious peg design allows easy viewing of tube contents
Use as a drying rack for glass plates or upside-down tubes
Pegs remain straight even after repeated autoclaving
Embossed alphanumeric identification for quick location and retrieval of tubes
Unique tear drop well design allows for better retention of tubes
Autoclavable
Still not finding the right product?
Explore all of our products under Peg Tube Rack
Choose from one of the most recent versions:
Already Own This Product?