Skip to Content
Merck
All Photos(3)

Key Documents

297887

Sigma-Aldrich

1-Octanol

anhydrous, ≥99%

Synonym(s):

Alcohol C8, Capryl alcohol, Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1697461
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
196 °C (lit.)
vapor pressure:
0.14 mmHg ( 25 °C)

grade

anhydrous

Quality Level

vapor density

4.5 (vs air)

vapor pressure

0.14 mmHg ( 25 °C)

Assay

≥99%

form

liquid

autoignition temp.

523 °F

expl. lim.

8 %

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.004%

refractive index

n20/D 1.429 (lit.)

bp

196 °C (lit.)

mp

−15 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCO

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Gene Information

mouse ... Chrna1(11435)

Looking for similar products? Visit Product Comparison Guide

General description

1-Octanol is a saturated fatty alcohol with potential application as a biofuel due to its diesel-like properties.

Application

1-Octanol may be used to prepare octanal via aerobic oxidation a polymer-supported 2,2,6,6-tetramethylpiperidine-N-oxyl as catalyst and Co(NO3)2 and Mn(NO3)2 as co-catalysts.

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Xiao et al.
Science advances, 3(9), e1701151-e1701151 (2017-09-21)
Structural colors enable the creation of a spectrum of nonfading colors without pigments, potentially replacing toxic metal oxides and conjugated organic pigments. However, significant challenges remain to achieve the contrast needed for a complete gamut of colors and a scalable
Ming Xiao et al.
Science advances, 5(9), eaax1254-eaax1254 (2019-09-27)
Surface segregation in binary colloidal mixtures offers a simple way to control both surface and bulk properties without affecting their bulk composition. Here, we combine experiments and coarse-grained molecular dynamics (CG-MD) simulations to delineate the effects of particle chemistry and
Polymer-supported nitroxyl radical catalyst for selective aerobic oxidation of primary alcohols to aldehydes
Gilhespy M, et al.
Chemical Communications (Cambridge, England), 8, 1085-1086 (2005)
Samantha J Carew et al.
Bio-protocol, 10(6), e3564-e3564 (2021-03-05)
Rats are highly social animals, and mainly communicate with one another in two ways: through ultrasonic vocalizations and pheromones. Most research on pheromones has been dedicated those regarding sexual behavior, but more recently pheromones which signal danger to conspecifics have
Clare E Hancock et al.
STAR protocols, 1(3), 100210-100210 (2020-12-31)
This protocol enables the quantification of odor-evoked calcium activity in mushroom body Kenyon cells of the Drosophila melanogaster brain at the single bouton level. We also present subsequent characterization of naive and learned odor representations in the context of olfactory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service