Skip to Content
Merck
All Photos(1)

Key Documents

T57630

Sigma-Aldrich

1-Tridecanol

97%

Synonym(s):

Tridecyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12OH
CAS Number:
Molecular Weight:
200.36
Beilstein:
1739991
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.9 (vs air)

Assay

97%

bp

155-156 °C/15 mmHg (lit.)

mp

29-34 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCO

InChI

1S/C13H28O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14/h14H,2-13H2,1H3

InChI key

XFRVVPUIAFSTFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Tridecanol is a primary alcohol which can undergo oxidative conversion to:
  • 2,2,2-trifluoroethyl tridecanoate.
  • Dodecanenitrile in the presence of molecular iodine in aq NH3.

It can also be used as a phase modifier for protonated trioctylamine in n-dodecane to study the speciation of vanadium (V) extracted from acidic sulfate media.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Speciation of vanadium (V) extracted from acidic sulfate media by trioctylamine in n-dodecane modified with 1-tridecanol.
Chagnes A, et al.
Hydrometallurgy, 104(1), 20-24 (2010)
Direct oxidative conversion of alcohols and amines to nitriles with molecular iodine and DIH in aq NH3.
Iida S and Togo H
Tetrahedron, 63(34), 8274-8281 (2007)
Facile oxidative conversion of alcohols to esters using molecular iodine.
Mori N and Togo H.
Tetrahedron, 61(24), 5915-5925 (2005)
D A Haydon et al.
Biochimica et biophysica acta, 863(2), 337-340 (1986-12-16)
The effects of 0.09 saturated solutions of the n-alkanols n-hexanol to n-tridecanol on the surface (compensation) potential of lipid monolayers have been examined. Actions on monolayers spread from pure egg phosphatidylcholine have been compared with effects on a system containing
D McKenzie et al.
British journal of pharmacology, 115(2), 275-282 (1995-05-01)
1. Completely isolated identified neurones from the right parietal ganglion of the pond snail Lymnaea stagnalis were studied under two-electrode voltage-clamp. Neuronal nicotinic acetylcholine receptor currents were studied at low acetylcholine (ACh) concentrations (< or = 200 nM). At these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service