Skip to Content
Merck
All Photos(3)

Documents

258768

Sigma-Aldrich

1-Octadecanol

ReagentPlus®, 99%

Synonym(s):

Octadecyl alcohol, Stearyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17OH
CAS Number:
Molecular Weight:
270.49
Beilstein:
1362907
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

9.3 (vs air)

vapor pressure

<0.01 mmHg ( 38 °C)

product line

ReagentPlus®

Assay

99%

autoignition temp.

842 °F

expl. lim.

~8 %

bp

170-171 °C/2 mmHg

mp

56-59 °C (lit.)

solubility

water: slightly soluble 0.001 g/L at 23 °C

density

0.812 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCO

InChI

1S/C18H38O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h19H,2-18H2,1H3

InChI key

GLDOVTGHNKAZLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mixed monolayers of 1-octadecanol and ethylene glycol monooctadecyl ether were studied to investigate their evaporation suppressing performance. The rate dependence of the collapse pressure for an octadecanolmonolayer using axisymmetric drop shape analysis has been investigated.

Application

1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

383.0 °F - closed cup

Flash Point(C)

195 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrical properties of natural rubber nanocomposites: effect of 1-octadecanol functionalization of carbon nanotubes.
Thomas PS, et al.
J. Mater. Sci., 47(7), 3344-3349 (2012)
Delaram Ahmadi et al.
Scientific reports, 10(1), 4082-4082 (2020-03-07)
Creams are multi-component semi-solid emulsions that find widespread utility across a wide range of pharmaceutical, cosmetic, and personal care products, and they also feature prominently in veterinary preparations and processed foodstuffs. The internal architectures of these systems, however, have to
Linards Klavins et al.
Foods (Basel, Switzerland), 9(5) (2020-05-10)
The outer-most layer of plant surface, the cuticle, consists of epi- and intra-cuticular wax. It protects the plant from dehydration, extreme temperatures and UV radiation, as well as attacks from pests such as molds and bacteria. Berry cuticular waxes are
Diana N H Tran et al.
The journal of physical chemistry. B, 117(13), 3603-3612 (2013-03-12)
Mixed monolayers of 1-octadecanol (C18OH) and ethylene glycol monooctadecyl ether (C18E1) were studied to assess their evaporation suppressing performance. An unexpected increase in performance and stability was found around the 0.5:0.5 bicomponent mixture and has been ascribed to a synergistic
Axisymmetric drop shape analysis as a film balance: rate dependence of the collapse pressure and molecular area at close packing of 1-octadecanol monolayers.
Kwok DY, et al.
Langmuir, 12(7), 1851-1859 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service