Skip to Content
Merck
All Photos(3)

Key Documents

P8001

Sigma-Aldrich

Sodium phenylpyruvate

powder

Synonym(s):

2-Oxo-3-phenylpropanoic acid, Phenylpyruvic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2COCOONa
CAS Number:
Molecular Weight:
186.14
Beilstein:
4770978
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

color

white to off-white

mp

>300 °C (lit.)

storage temp.

2-8°C

SMILES string

[Na+].[O-]C(=O)C(=O)Cc1ccccc1

InChI

1S/C9H8O3.Na/c10-8(9(11)12)6-7-4-2-1-3-5-7;/h1-5H,6H2,(H,11,12);/q;+1/p-1

InChI key

MQGYVGKMCRDEAF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Wang et al.
Frontiers in microbiology, 10, 2357-2357 (2019-11-05)
Endogenous hydrogen sulfide (H2S), which is primarily generated by 3-mercaptopyruvate sulfurtransferase (3-MST) in Escherichia coli (E. coli) under aerobic conditions, renders bacteria highly resistant to oxidative stress. However, the biosynthetic pathway and physiological role of this gas under anaerobic conditions
Cristiana M L Di Giuro et al.
PloS one, 8(7), e68932-e68932 (2013-08-13)
(S)-Hydroxymandelate synthase (Hms) is a nonheme Fe(II) dependent dioxygenase that catalyzes the oxidation of 4-hydroxyphenylpyruvate to (S)-4-hydroxymandelate by molecular oxygen. In this work, the substrate promiscuity of Hms is characterized in order to assess its potential for the biosynthesis of
William M Moe et al.
Environmental science & technology, 52(16), 9311-9319 (2018-07-26)
In situ bioremediation practices that include subsurface addition of fermentable electron donors to stimulate reductive dechlorination by anaerobic bacteria have become widely employed to combat chlorinated solvent contamination in groundwater. At a contaminated site located near Baton Rouge, Louisiana (USA)
Hajer Ouertatani-Sakouhi et al.
Journal of biomolecular screening, 15(4), 347-358 (2010-03-17)
Macrophage migration inhibitory factor (MIF) is a major mediator of innate immunity and inflammation and presents a potential therapeutic target for various inflammatory, infectious, and autoimmune diseases, including cancer. Although a number of inhibitors have been identified and designed based
Tapan Kanti Paine et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(21), 6073-6081 (2007-05-01)
Iron(II)-phenylpyruvate complexes of tetradentate tris(6-methyl-2-pyridylmethyl)amine (6-Me3-TPA) and tridentate benzyl bis(2-quinolinylmethyl)amine (Bn-BQA) were prepared to gain insight into C-C bond cleavage catalyzed by dioxygenase enzymes. The complexes we have prepared and characterized are [Fe(6-Me3-tpa)(prv)][BPh4] (1), [Fe2(6-Me3-tpa)2(pp)][(BPh4)2] (2), and [Fe2(6-Me3-tpa)2(2'-NO2-pp)][(BPh4)2] (3), [Fe(6-Me3-tpa)(pp-Me)][BPh4]

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service