Skip to Content
Merck
All Photos(1)

Key Documents

P7251

Sigma-Aldrich

3-Phenyllactic acid

≥98%

Synonym(s):

α-Hydroxyhydrocinnamic acid, β-Phenyllactic acid, 2-Hydroxy-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(OH)COOH
CAS Number:
Molecular Weight:
166.17
Beilstein:
2209791
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

SMILES string

OC(Cc1ccccc1)C(O)=O

InChI

1S/C9H10O3/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)

InChI key

VOXXWSYKYCBWHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Phenyllactic acid can be used as a reactant to prepare:
  • O




  • -Acetyl-3-phenyllactic acid by reacting with acetic anhydride in the presence of pyridine.
  • Ethyl 2-oxo-1-(phenylmethyl)-2-[(phenylmethyl)amino]ethyl carbonate by one pot reaction with 2-ethoxy-1-(ethoxycarbonyl)-1,2-dihydroquinoline (EEDQ) and aniline.
  • Phenylmethyl-1,3-dioxolane-2,4-dione by condensation reaction with trichloromethyl chloroformate (diphosgene) in the presence of activated charcoal.

Reagent involved in biological studies of:
  • Enantioselectivity of lipase in transesterification
  • Oxidation by glycolate oxidase and catalase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mattia Quattrini et al.
International journal of food microbiology, 302, 8-14 (2018-09-18)
Fungal spoilage of bread remains an unsolved issue in bread making. This work aims to identify alternative strategies to conventional preservatives in order to prevent or delay fungal spoilage of bread. The minimum inhibitory concentration (MIC) of bacterial metabolites and
Tiago de Melo Nazareth et al.
Toxins, 12(1) (2020-01-08)
Fungal spoilage is an important issue for the food industry, leading to food sensory defects, food waste, economic losses and public health concern through the production of mycotoxins. Concomitantly, the search for safer natural products has gained importance since consumers
Design and structure-activity relationships of potent and selective inhibitors of undecaprenyl pyrophosphate synthase (UPPS): tetramic, tetronic acids and dihydropyridin-2-ones
Peukert S, et al.
Bioorganic & medicinal chemistry letters, 18(6), 1840-1844 (2008)
C N Sarkissian et al.
Analytical biochemistry, 280(2), 242-249 (2000-05-03)
Phenylketonuria (PKU) (OMIM 261600) is the first Mendelian disease to have an identified chemical cause of impaired cognitive development. The disease is accompanied by hyperphenylalaninemia (HPA) and elevated levels of phenylalanine metabolites (phenylacetate (PAA), phenyllactate (PLA), and phenylpyruvate (PPA)) in
Use of 2-ethoxy-1-(ethoxycarbonyl)-1, 2-dihydroquinoline as a convenient reagent for the selective protection or derivatization of 2-hydroxycarboxylic acids
Hyun Myung Ho, et al.
Tetrahedron Letters, 40(17), 3435-3438 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service