Skip to Content
Merck
All Photos(1)

Key Documents

677175

Sigma-Aldrich

Pyruvic-1-13C acid

≥99 atom % 13C, ≥99% (CP)

Synonym(s):

α-Ketopropionic acid-1-13C, 2-Oxopropionic acid-1-13C, 13C Labeled pyruvic acid, Pyruvic acid-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO13CO2H
CAS Number:
Molecular Weight:
89.05
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥99% (CP)

form

liquid

bp

165 °C

mp

11-12 °C

density

1.281 g/mL at 25 °C

mass shift

M+1

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(=O)[13C](O)=O

InChI

1S/C3H4O3/c1-2(4)3(5)6/h1H3,(H,5,6)/i3+1

InChI key

LCTONWCANYUPML-LBPDFUHNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyruvic-1-13C is isotopic labeled precursor of pyruvic acid.

Application

Hyperpolarized 13C-labeled pyruvate and its metabolite lactate has been used to image real time metabolic flux in solid tumors and assess tumor response to chemotherapy, radiation, and agents targeting mTOR.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mette Hauge Lauritzen et al.
NMR in biomedicine, 26(11), 1496-1500 (2013-06-25)
A change in myocardial metabolism is a known effect of several diseases. MRS with hyperpolarized (13)C-labelled pyruvate is a technique capable of detecting changes in myocardial pyruvate metabolism, and has proven to be useful for the evaluation of myocardial ischaemia
Prasanta Dutta et al.
Cancer research, 73(14), 4190-4195 (2013-06-01)
Hyperpolarized (13)C magnetic resonance spectroscopy provides a unique opportunity to detect real-time metabolic fluxes as a means to measure metabolic treatment responses in vivo. Here, we show that pharmacologic inhibition of lactate dehydrogenase-A suppressed the conversion of hyperpolarized (13)C-pyruvate to
Ilwoo Park et al.
Journal of magnetic resonance imaging : JMRI, 33(6), 1284-1290 (2011-05-19)
To demonstrate the feasibility of using DNP hyperpolarized [1-(13)C]-pyruvate to measure early response to temozolomide (TMZ) therapy using an orthotopic human glioblastoma xenograft model. Twenty athymic rats with intracranial implantation of human glioblastoma cells were divided into two groups: one
Albert P Chen et al.
Magnetic resonance in medicine, 73(6), 2087-2093 (2014-07-22)
Hyperpolarized [1-(13) C]lactate in solution may be a clinically relevant and safe substrate for real time MR investigations of key metabolic pathways. The potential of using hyperpolarized [1-(13) C]lactate for magnetic resonance studies of cardiac metabolism in vivo was explored.

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service