Skip to Content
Merck
All Photos(3)

Key Documents

P0677

Sigma-Aldrich

Potassium tellurite

≥90%

Synonym(s):

Dipotassium tellurite, Potassium tellurate, Potassium tellurium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2TeO3
CAS Number:
Molecular Weight:
253.79
EC Number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥90%

form

powder

SMILES string

O=[Te](O)O

InChI

1S/H2O3Te/c1-4(2)3/h(H2,1,2,3)

InChI key

SITVSCPRJNYAGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium tellurite is a hygroscopic granular white powder. It decomposes at 460-470oC.

Application

Potassium tellurite along with agar may be used as a component of a selective medium for growth of bacteria.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of potassium tellurite for rapid-drug susceptibility testing of Mycobacterium avium complex
Afgani B and Fujiyama D
Journal of Investigative Medicine, 49, 292-296 null
Giulio Zanaroli et al.
Research in microbiology, 153(6), 353-360 (2002-09-18)
This study shows that the oxyanion tellurite TeO3(2-) can be used as a tool to detect and quantify the release in soil microcosms of Pseudomonas pseudoalcaligenes KF707, a strain spontaneously resistant to tellurite with a minimal inhibitory concentration (MIC) of
Francesca Romana Cianfanelli et al.
BMC microbiology, 20(1), 129-129 (2020-05-26)
Gene editing is key for elucidating gene function. Traditional methods, such as consecutive single-crossovers, have been widely used to modify bacterial genomes. However, cumbersome cloning and limited efficiency of negative selection often make this method slower than other methods such
Elkin Jahir Florez Salamanca et al.
Biofouling, 36(10), 1196-1209 (2020-12-23)
Biofilms were developed from human saliva on bovine enamel discs in four experimental conditions to investigate dental caries development: feast and famine (M1), abundance and scarcity (M2), three meals daily (M3), and three meals plus two snacks daily (M4). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service