M90003
trans,trans-Muconic acid
95%
Synonym(s):
trans,trans-1,3-Butadiene-1,4-dicarboxylic acid, trans,trans-2,4-Hexadienedioic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO2CCH=CHCH=CHCO2H
CAS Number:
Molecular Weight:
142.11
Beilstein:
1722248
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
powder
bp
320 °C (lit.)
mp
290 °C (dec.) (lit.)
SMILES string
OC(=O)\C=C\C=C\C(O)=O
InChI
1S/C6H6O4/c7-5(8)3-1-2-4-6(9)10/h1-4H,(H,7,8)(H,9,10)/b3-1+,4-2+
InChI key
TXXHDPDFNKHHGW-ZPUQHVIOSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
trans,trans-Muconic acid is a polyunsaturated dicarboxylic acid characterized by its conjugated double bonds and reactive dicarboxylic groups. It serves primarily as a key precursor to monomers in the synthesis of polyamides and polyesters.
Application
trans,trans-Muconic acid can be used as a:
- Reactant to produce aromatic monomers via Diels-Aldercycloaddition.
- Biomarker for benzene exposure.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nancy B Hopf et al.
International archives of occupational and environmental health, 85(3), 261-271 (2011-06-15)
Characterize ethylbenzene and xylene air concentrations, and explore the biological exposure markers (urinary t,t-muconic acid (t,t-MA) and unmetabolized toluene) among petroleum workers offshore. Offshore workers have increased health risks due to simultaneous exposures to several hydrocarbons present in crude oil.
Occupational exposure to benzene and chromosomal structural aberrations in the sperm of Chinese men.
Francesco Marchetti et al.
Environmental health perspectives, 120(2), 229-234 (2011-11-17)
Benzene is an industrial chemical that causes blood disorders, including acute myeloid leukemia. We previously reported that occupational exposures near the U.S. Occupational Safety and Health Administration permissible exposure limit (8 hr) of 1 ppm was associated with sperm aneuploidy.
G Scherer et al.
Journal of chromatography. B, Biomedical sciences and applications, 717(1-2), 179-199 (1998-12-01)
Benzene is an important industrial chemical and, due to its occurrence in mineral oil and its formation in many combustion processes, a widespread environmental pollutant. Since benzene is hematoxic and has been classified as a human carcinogen, monitoring and control
A Bartczak et al.
Journal of toxicology and environmental health, 42(3), 245-258 (1994-07-01)
Muconic acid (MA) is a urinary metabolite of benzene and has been used as a biomarker of exposure to benzene in humans exposed to levels as low as 1 ppm. We have modified a high-pressure liquid chromatography (HPLC) based assay
Sirirat Chanvaivit et al.
Mutation research, 626(1-2), 79-87 (2006-11-11)
Exposure to benzene can cause DNA damage and the subsequent development of cancer. In this study, study subjects were 31 laboratory workers at a petrochemical factory and 31 gasoline service attendants. Control subjects were 34 workers from a mail sorting
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service