Skip to Content
Merck
All Photos(1)

Key Documents

328006

Sigma-Aldrich

2-Methylhippuric acid

98%

Synonym(s):

N-(2-Methylbenzoyl)glycine, o-Toluric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CONHCH2CO2H
CAS Number:
Molecular Weight:
193.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

powder

mp

161-164 °C (lit.)

functional group

amide
carboxylic acid

SMILES string

Cc1ccccc1C(=O)NCC(O)=O

InChI

1S/C10H11NO3/c1-7-4-2-3-5-8(7)10(14)11-6-9(12)13/h2-5H,6H2,1H3,(H,11,14)(H,12,13)

InChI key

YOEBAVRJHRCKRE-UHFFFAOYSA-N

Related Categories

General description

2-Methylhippuric acid is also known as o-methylhippuric acid and its determination in urine by gas chromatography was reported.[1] It is the urinary metabolic biomarker for the diagnosis of human Hepatitis B virus.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sirikool Thamnium et al.
Scientific reports, 13(1), 14102-14102 (2023-08-30)
Asiatic acid (AA) and asiaticoside, pentacyclic triterpenoid compounds derived from Centella asiatica, are known for their biological effects in promoting type I collagen synthesis and inducing osteogenesis of stem cells. However, their applications in regenerative medicine are limited due to
Aihua Zhang et al.
PloS one, 8(5), e64381-e64381 (2013-05-23)
Hepatitis B virus (HBV) is the fatal consequence of chronic hepatitis, and lack of biomarkers has been a long standing bottleneck in the clinical diagnosis. Metabolomics concerns with comprehensive analysis of small molecules and provides a powerful approach to discover
M Ogata et al.
International archives of occupational and environmental health, 61(1-2), 131-140 (1988-01-01)
An attempt was made to establish a method for the direct determination of urinary concentrations of creatinine, hippuric acid, methylhippuric acid and mandelic acid by automated high performance liquid chromatography (HPLC). Urine was diluted with distilled water or mobile phase
M Ogata et al.
International archives of occupational and environmental health, 58(2), 121-129 (1986-01-01)
A new method for the direct determination of hippuric acid (HA) and o-, m- and p-methylhippuric acids (MHAs) in the urine, metabolites of toluene and o-, m- and p-xylenes by high performance liquid chromatography (HPLC) is described. A stainless-steel column
Simultaneous determination of hippuric acid and o-, m- and p-methylhippuric acids in urine by high-performance liquid chromatography.
T Sakai et al.
Journal of chromatography, 276(1), 182-188 (1983-08-12)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service