H29404
1-(2-Hydroxyethyl)pyrrolidine
97%
Synonym(s):
2-Pyrrolidinoethanol, Pyrrolidine-1-ethanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
Beilstein:
102976
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.4713
bp
79-81 °C/13 mmHg (lit.)
density
0.985 g/mL at 25 °C (lit.)
SMILES string
OCCN1CCCC1
InChI
1S/C6H13NO/c8-6-5-7-3-1-2-4-7/h8H,1-6H2
InChI key
XBRDBODLCHKXHI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
132.8 °F - closed cup
Flash Point(C)
56 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
C Giachetti et al.
European journal of drug metabolism and pharmacokinetics, 21(3), 261-268 (1996-07-01)
N-(2-hydroxyethyl)-pyrrolidine (HEP, Epolamine) is a strong base used to salify organic acids of pharmaceutical interest in order to improve their solubility in water. Diclofenac-HEP (Flector) is the first example of an epolamine salt of a drug. In this study, [14C]-HEP
Joseph Gimbel et al.
The Physician and sportsmedicine, 39(1), 11-18 (2011-03-08)
The diclofenac epolamine topical patch 1.3% (DETP) is indicated for topical treatment of acute pain due to minor strains, sprains, and contusions. This exploratory, multicenter, open-label study evaluated effectiveness and safety of DETP in patients with acute pain due to
Adamo Fini et al.
Pharmaceutics, 4(3), 413-429 (2012-01-01)
The following bases: monoethylamine (EtA), diethylamine (DEtA), triethylamine (TEtA), monoethanolamine (MEA), diethanolamine (DEA), triethanolamine (TEA), pyrrolidine (Py), piperidine (Pp), morpholine (M), piperazine (Pz) and their N-2-hydroxyethyl (HE) analogs were employed to prepare 14 diclofenac salts. The salts were re-crystallized from
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service