Skip to Content
Merck
All Photos(1)

Documents

272264

Sigma-Aldrich

1-(3-Aminopropyl)imidazole

≥97%

Synonym(s):

1-(3-Aminopropanyl)imidazole, 3-(1-Imidazolyl)propylamine, 3-(1H-Imidazol-1-yl)propylamine, 3-(Imidazol-1-yl)propylamine, N-(3-Aminopropyl)imidazole, N-[3-(1H-Imidazol-1-yl)propyl]amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N3
CAS Number:
Molecular Weight:
125.17
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

refractive index

n20/D 1.519 (lit.)

density

1.049 g/mL at 25 °C (lit.)

SMILES string

NCCCn1ccnc1

InChI

1S/C6H11N3/c7-2-1-4-9-5-3-8-6-9/h3,5-6H,1-2,4,7H2

InChI key

KDHWOCLBMVSZPG-UHFFFAOYSA-N

Application

1-(3-Aminopropyl)imidazole was used in the synthesis of pH-sensitive polyaspartamide derivatives. It was also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution by graft reaction.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

309.2 °F

Flash Point(C)

154 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kwangwon Seo et al.
Journal of nanoscience and nanotechnology, 10(10), 6986-6991 (2010-12-09)
The effect of hydrophobic octadecyl groups on pH-dependent aggregation behavior of polyaspartammide derivatives was studied. A series of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution were synthesized through a successive graft reaction of octaceylamine, O-(2-aminoethyl)-O'-methylpolyethylene glycol, and
Taehoon Sim et al.
Pharmaceutics, 12(9) (2020-09-06)
Combination therapy is considered to be a promising strategy for improving the therapeutic efficiency of cancer treatment. In this study, an on-demand pH-sensitive nanocluster (NC) system was prepared by the encapsulation of gold nanorods (AuNR) and doxorubicin (DOX) by a
Abootaleb Sedighi et al.
Talanta, 186, 568-575 (2018-05-23)
Several solution-based methods have recently been adapted for use in paper substrates for enzymatic amplification to increase the number of copies of DNA sequences. There is limited information available about the impact of a paper matrix on DNA amplification by
Xiaodan Li et al.
Journal of controlled release : official journal of the Controlled Release Society, 260, 12-21 (2017-05-22)
Photodynamic therapy (PDT) efficacy is limited by the very short half-life and limited diffusion radius of singlet oxygen (
Kajari Chatterjee et al.
ACS omega, 5(27), 16681-16689 (2020-07-21)
Our society is critically dependent on lithium-ion batteries (LIBs) as a power source for portable electronic gadgets. One of the major problems with these batteries is the degradation of the materials inside them. In addition to the reduced cell life

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service